Skip to Content
Merck
All Photos(1)

Documents

SML2401

Sigma-Aldrich

Imeglimin hydrochloride

≥98% (HPLC), powder, antidiabetic agent

Synonym(s):

(6R)-1,6-dihydro-N,N,6-trimethyl-1,3,5-Triazine-2,4-diamine Hydrochloride,, (6R)-3,6-Dihydro-N2,N2,6-trimethyl-1,3,5-triazine-2,4-diamine, EMD 387008 Hydrochloride, EMD-387008 Hydrochloride, R-(4-Imino-6-methyl-1,4,5,6-tetrahydro-[1,3,5]trazin-2-yl)dimethylamine Hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13N5 · xHCl
CAS Number:
Molecular Weight:
155.20 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

product name

Imeglimin hydrochloride, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C6H13N5.ClH/c1-4-8-5(7)10-6(9-4)11(2)3;/h4H,1-3H3,(H3,7,8,9,10);1H/t4-;/m1./s1

InChI key

UXHLCYMTNMEXKZ-PGMHMLKASA-N

Biochem/physiol Actions

Imeglimin is a first-in-class oral glucose-lowering agent whose mechanism of action targets mitochondrial bioenergetics, resulting in increased insulin secretion in response to glucose, improved glucose uptake by skeletal muscle, and suppression of gluconeogenesis. Imeglimin has been shown to decrease reactive oxygen species overproduction and delay mPTP opening, preventing cell death during oxidative stress, protecting beta-cells. In an animal model, it has been shown to reduce metabolic syndrome-related diabetic cardiomyopathy.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P577 Short- and long-term imeglimin treatment reduces metabolic syndrome-related diabetic cardiomyopathy
Lachaux M, Hamzaoui M, Nicol L, Fouqueray P, Bolze S, Hallakou-Bozec S, Richard V, Mulder P
European Heart Journal, 38 (2017)
D Detaille et al.
Cell death discovery, 2, 15072-15072 (2016-08-24)
Imeglimin is the first in a new class of oral glucose-lowering agents, having recently completed its phase 2b trial. As Imeglimin did show a full prevention of β-cell apoptosis, and since angiopathy represents a major complication of diabetes, we studied

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service