Skip to Content
Merck
All Photos(1)

Documents

SML2396

Sigma-Aldrich

α-DHED

≥98% (HPLC)

Synonym(s):

10ß,17a-Dihydroxyestra-1,4-dien-3-one, alpha-DHED

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24O3
CAS Number:
Molecular Weight:
288.38
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O[C@@]12C(CC[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@H]4O)=CC(C=C1)=O

Biochem/physiol Actions

α-DHED is a bioprecursor prodrug of 17α-estradiol ((Alfatradiol)) that results in 17α-estradiol being produced selectively in the brain with little production in the periphery. α-DHED has no estrogenic activity itself with little affinity for either ERα and ERβ estrogen receptors. 17α-estradiol (αE2) has been proposed to have potential roles in various neurodegenerative disorders including neuroprotection of the brain and the prevention/treatment of Alzheimer′s disease, but peripheral side-effects on estrogen-sensitive organs have been problematic. α-DHED, a prodrug without estrogenic activity, could help alleviate those concerns.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katalin Prokai-Tatrai et al.
ACS chemical neuroscience, 9(11), 2528-2533 (2018-05-31)
Uterotrophic effect of 17α-estradiol, the C17 epimer of the main human estrogen 17β-estradiol, was shown to manifest in animal models at doses lower than those necessary for central outcome raising concerns about its potential to treat maladies of the central

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service