Skip to Content
Merck
All Photos(1)

Key Documents

SML2350

Sigma-Aldrich

Ferroquine

≥98% (HPLC)

Synonym(s):

1-[[(7-Chloro-4-quinolinyl)amino]methyl]-2-[(dimethylamino)methyl]-ferrocene, 7-Chloro-4-(((2-((dimethylamino)methyl)ferrocenyl)methyl)amino)quinoline, Ferrochloroquine, SSR 97193

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$168.00
25 MG
$672.00

$168.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$168.00
25 MG
$672.00

About This Item

Empirical Formula (Hill Notation):
C23H24ClFeN3
CAS Number:
Molecular Weight:
433.75
UNSPSC Code:
12352200
NACRES:
NA.77

$168.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

yellow to brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CN(C)C[C]1[C]C[C][C]1CNC2=C(C=CC(Cl)=C3)C3=NC=C2.[C]4[C][C][C]C4.[Fe]

Biochem/physiol Actions

Antimalarial effective against chloroquine-resistant Plasmodium falciparum
Ferroquine (SSR 97193) is an antimalarial chloroquine analog effective against chloroquine-resistant Plasmodium falciparum. Ferroquine is thought to act by producing oxidative stress and by inhibiting the formation of hemozoin, produced by Malaria parasites as a disposal product of heme, which would otherwise be toxic to the Plasmodium cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jana Held et al.
The Lancet. Infectious diseases, 15(12), 1409-1419 (2015-09-08)
Artemisinin-based combination therapies (ACTs) are the recommended first-line treatment for uncomplicated Plasmodium falciparum malaria. Ferroquine is a new combination partner for fast-acting ACTs such as artesunate. We aimed to assess different doses of ferroquine in combination with artesunate against uncomplicated
Waseem A Wani et al.
European journal of medicinal chemistry, 101, 534-551 (2015-07-21)
Malaria has been teasing human populations from a long time. Presently, several classes of antimalarial drugs are available in market, but the issues of toxicity, lower efficacy and the resistance by malarial parasites have decreased their overall therapeutic indices. Thus
Artem Kondratskyi et al.
Scientific reports, 7(1), 15896-15896 (2017-11-23)
Despite the tremendous progress in medicine, cancer remains one of the most serious global health problems awaiting new effective therapies. Here we present ferroquine (FQ), the next generation antimalarial drug, as a promising candidate for repositioning as cancer therapeutics. We

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service