Skip to Content
Merck
All Photos(1)

Key Documents

SML2309

Sigma-Aldrich

Oxaphenamide

≥98% (HPLC)

Synonym(s):

2-Hydroxy-N-(4-hydroxyphenyl)benzamide, 4′-Hydroxysalicylanilide, Osalmide, Oxaphenamide, p-hydroxyphenylsalicylamide

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$128.00

$128.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$128.00

About This Item

Empirical Formula (Hill Notation):
C13H11NO3
CAS Number:
Molecular Weight:
229.23
EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$128.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N(c2ccc(cc2)O)C(=O)c1c(cccc1)O

InChI

1S/C13H11NO3/c15-10-7-5-9(6-8-10)14-13(17)11-3-1-2-4-12(11)16/h1-8,15-16H,(H,14,17)

InChI key

LGCMKPRGGJRYGM-UHFFFAOYSA-N

Biochem/physiol Actions

Osalmid is a potent inhibitor of ribonucleotide reductase small subunit M2 (RRM2) that potently inhibits HBV DNA and cccDNA synthesis in HepG2.2.15 cells. Osalmid inhibits RR (ribonucleotide reductase) activity in vivo, and exhibits synergistic effect with lamivudine on inactivation mutant HBV strain. Osalmid is a biliation activator (choleretic agent), but a mechanism of this activity remains unknown.
potent inhibitor of ribonucleotide reductase small subunit M2 (RRM2) that potently inhibits HBV DNA and cccDNA synthesis; choleretic agent

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service