Skip to Content
Merck
All Photos(1)

Key Documents

SML2264

Sigma-Aldrich

Col003

≥98% (HPLC)

Synonym(s):

2-Hydroxy-3-nitro-5-(phenylmethyl)benzaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$174.00
25 MG
$555.00

$174.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$174.00
25 MG
$555.00

About This Item

Empirical Formula (Hill Notation):
C14H11NO4
CAS Number:
Molecular Weight:
257.24
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$174.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

yellow to orange

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC1=C(C=C(C=C1C=O)CC2=CC=CC=C2)[N+]([O-])=O

InChI

1S/C14H11NO4/c16-9-12-7-11(6-10-4-2-1-3-5-10)8-13(14(12)17)15(18)19/h1-5,7-9,17H,6H2

InChI key

RKJXVCLOGNLZOI-UHFFFAOYSA-N

Biochem/physiol Actions

Col003 is a cell penetrant, selective and potent inhibitor of the interaction of Hsp47 with collagen. It binds to the collagen binding site on Hsp47. Col003 inhibits a collagen secretion by destabilization the collagen triple helix. Additionally Col003 weakly inhibits interaction of collagen with prolyl 4-hydroxylase (P4H).
cell penetrant, selective and potent inhibitor of the interaction of Hsp47 with collagen

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shinya Ito et al.
The Journal of biological chemistry, 292(49), 20076-20085 (2017-10-14)
Fibrosis can disrupt tissue structure and integrity and impair organ function. Fibrosis is characterized by abnormal collagen accumulation in the extracellular matrix. Pharmacological inhibition of collagen secretion therefore represents a promising strategy for the management of fibrotic disorders, such as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service