Skip to Content
Merck
All Photos(1)

Key Documents

SML2195

Sigma-Aldrich

AZD6482

≥98% (HPLC)

Synonym(s):

(-) 2-[1-(7-methyl-2-(morpholin-4-yl)-4-oxo-4H-pyrido[1,2-a]pyrimidin-9-yl)ethylamino]benzoic acid, 2-[[(1R)-1-[7-Methyl-2-(4-morpholinyl)-4-oxo-4H-pyrido[1,2-a]pyrimidin-9-yl]ethyl]amino]-benzoic acid, AZD 6482

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$166.00
25 MG
$665.00

$166.00


Estimated to ship on14 April 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$166.00
25 MG
$665.00

About This Item

Empirical Formula (Hill Notation):
C22H24N4O4
CAS Number:
Molecular Weight:
408.45
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$166.00


Estimated to ship on14 April 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[n]21c(nc(c[c]2=O)N4CCOCC4)c(cc(c1)C)[C@H](Nc3c(cccc3)C(=O)O)C

InChI

1S/C22H24N4O4/c1-14-11-17(15(2)23-18-6-4-3-5-16(18)22(28)29)21-24-19(12-20(27)26(21)13-14)25-7-9-30-10-8-25/h3-6,11-13,15,23H,7-10H2,1-2H3,(H,28,29)/t15-/m1/s1

InChI key

IRTDIKMSKMREGO-OAHLLOKOSA-N

Biochem/physiol Actions

AZD6482 is an isoform-selective and potent ATP competitive inhibitor of PI3Kb. AZD6482 produces an antithrombotic effect without an increased bleeding time or blood loss in humans and in dogs.
isoform-selective and potent ATP competitive inhibitor of PI3Kb

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Britta Weigelt et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 19(13), 3533-3544 (2013-05-16)
Endometrioid endometrial cancers (EEC) frequently harbor coexisting mutations in phosphoinositide 3-kinase (PI3K) pathway genes, including PTEN, PIK3CA, PIK3R1, and KRAS. We sought to define the genetic determinants of PI3K pathway inhibitor response in EEC cells, and whether PTEN-mutant EEC cell
S Nylander et al.
Journal of thrombosis and haemostasis : JTH, 10(10), 2127-2136 (2012-08-22)
Based on in vitro and animal data, PI3Kβ is given an important role in platelet adhesion and aggregation but its role in insulin signaling is unclear. To strengthen the PI3Kβ target validation using the novel, short-acting inhibitor AZD6482. AZD6482 is
Chen Zhang et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 37(11), 14831-14839 (2016-09-19)
Breast cancer is one of the leading causes of death for women worldwide. Among various subtypes of breast cancer, human epidermal growth factor receptor 2 (HER2)-positive and phosphatase and tensin homolog (PTEN) loss breast cancer is a cause of great
Hon Yan K Yip et al.
Molecular cell, 80(2), 279-295 (2020-10-17)
The PTEN tumor suppressor controls cell death and survival by regulating functions of various molecular targets. While the role of PTEN lipid-phosphatase activity on PtdIns(3,4,5)P3 and inhibition of PI3K pathway is well characterized, the biological relevance of PTEN protein-phosphatase activity
Sameer S Chopra et al.
Cell systems, 10(1), 66-81 (2019-12-10)
Frequent mutation of PI3K/AKT/mTOR signaling pathway genes in human cancers has stimulated large investments in targeted drugs but clinical successes are rare. As a result, many cancers with high PI3K pathway activity, such as triple-negative breast cancer (TNBC), are treated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service