Skip to Content
Merck
All Photos(1)

Key Documents

SML2184

Sigma-Aldrich

Kevetrin hydrochloride

≥98% (HPLC)

Synonym(s):

Kevetrin hydrochloride, 4-Isothioureidobutyronitrile hydrochloride, Carbamimidothioic acid 3-cyanopropyl ester hydrochloride, NSC 525990 HCl

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$148.00
25 MG
$472.00

$148.00


Estimated to ship on29 April 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$148.00
25 MG
$472.00

About This Item

Empirical Formula (Hill Notation):
C5H9N3S · HCl
CAS Number:
Molecular Weight:
179.67
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$148.00


Estimated to ship on29 April 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

NC(SCCCC#N)=N.Cl

InChI

1S/C5H9N3S.ClH/c6-3-1-2-4-9-5(7)8;/h1-2,4H2,(H3,7,8);1H

InChI key

NCXJZJFDQMKRKM-UHFFFAOYSA-N

Biochem/physiol Actions

Kevetrin is an activator/stabilizer of wild type p53 tumor suppressor protein in human lung carcinoma and ovarian cancer cells that induces cell cycle arrest and apoptosis. Additionally Kevetrin down-regulates oncogenic mutant p53 in OVCAR-3 ovarian cancer cells. Kevetrin inhibits tumor growth in xenograft model of ovarian cancer.
wild type p53 stabilizer

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service