Skip to Content
Merck
All Photos(1)

Key Documents

SML2164

Sigma-Aldrich

Bepristat 1a

≥98% (HPLC)

Synonym(s):

1-[(3-Chloro-4-hydroxyphenyl)methyl]-N-ethyl-N-methyl-4-(2-phenoxyethyl)-4-piperidinecarboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$150.00
25 MG
$587.00

$150.00


Available to ship on10 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 MG
$150.00
25 MG
$587.00

About This Item

Empirical Formula (Hill Notation):
C24H31ClN2O3
CAS Number:
Molecular Weight:
430.97
UNSPSC Code:
12352200
NACRES:
NA.77

$150.00


Available to ship on10 April 2025Details


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC1=C(O)C=CC(CN2CCC(C(N(CC)C)=O)(CCOC3=CC=CC=C3)CC2)=C1

InChI key

AYJKPBTXOIHUFT-UHFFFAOYSA-N

Biochem/physiol Actions

Bepristat 1a is a selective reversible inhibitor of protein disulfide isomerase (PDI), an enzyme in the endoplasmic reticulum that catalyzes disulfide bond breakage and reformation to catalyze protein folding. Unlike most PDI inhibitors, Bepristat 1a binds at the substrate-binding site, rather than the catalytic site. It blocked PDI activity with an IC50 value of 700 nM. PDI is up-regulated in several cancers, has been implicated in neurodegenerative processes, and plays an important role in thrombus formation. Bepristat 1a potently inhibited platelet aggregation and thrombus formation in vivo.
Selective reversible inhibitor of protein disulfide isomerase (PDI)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roelof H Bekendam et al.
Nature communications, 7, 12579-12579 (2016-08-31)
Protein disulfide isomerase (PDI) is an oxidoreductase essential for folding proteins in the endoplasmic reticulum. The domain structure of PDI is a-b-b'-x-a', wherein the thioredoxin-like a and a' domains mediate disulfide bond shuffling and b and b' domains are substrate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service