Skip to Content
Merck
All Photos(1)

Key Documents

SML2158

Sigma-Aldrich

VX-680

≥98% (HPLC)

Synonym(s):

MK 0457, MK-0457, MK0457, N-[4-[[4-(4-Methyl-1-piperazinyl)-6-[(5-methyl-1H-pyrazol-3-yl)amino]-2-pyrimidinyl]thio]phenyl]cyclopropanecarboxamide, Tozasertib, VX 680, VX680

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$117.00

$117.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$117.00

About This Item

Empirical Formula (Hill Notation):
C23H28N8OS
CAS Number:
Molecular Weight:
464.59
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$117.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1CC1)NC(C=C2)=CC=C2SC3=NC(N4CCN(C)CC4)=CC(NC5=NNC(C)=C5)=N3

InChI

1S/C23H28N8OS/c1-15-13-20(29-28-15)25-19-14-21(31-11-9-30(2)10-12-31)27-23(26-19)33-18-7-5-17(6-8-18)24-22(32)16-3-4-16/h5-8,13-14,16H,3-4,9-12H2,1-2H3,(H,24,32)(H2,25,26,27,28,29)

InChI key

GCIKSSRWRFVXBI-UHFFFAOYSA-N

Biochem/physiol Actions

ATP site-targeting potent aurora kinase and Abl inhibitor with in vitro and in vivo anticancer efficacy.
VX-680 is an ATP site-targeting potent aurora kinase inhibitor (Aurara A/B/C Ki(app) = 0.6/18/4.6 nM) that affects FLT3, BCR-Abl, BCR-Abl (T315I), Lck, ITK, Src, and Fyn only at higher concentrations (Ki(app) = 30, 30, 42, 80, 220, 350, 520 nM, respectively) and exhibits little inhibitory potency toward 52 other kinases (Ki(app) >1 μM). VX-680 exhibits potent antiproliferation activity in a wide variety of cancer cultures (IC50 from 15 to 113 nM) as a result of cell cycle arrest and apoptosis induction, as well as causes tumor retardation (by 98% on day 13; 75 mg/kg b.i.d i.p.; HL-60 in mice) and regression (2 mg/kg/h 3 d/wk i.v. infusion; HCT116 in rats) in vivo. Crystallography data reveal a tight association of VX-680 with a hydrophobic pocket present only in a closed, inactive kinase conformation, which forms the basis of its selectivity profile, including its activity toward wild-type and the Imatinib-resistant (T315I) Abl.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G M T Cheetham et al.
Cancer letters, 251(2), 323-329 (2007-01-24)
The small molecule inhibitor of the Aurora-family of protein kinases VX-680 or MK-0457, demonstrates potent anti-cancer activity in multiple in vivo models and has recently entered phase II clinical trials. Although VX-680 shows a high degree of enzyme selectivity against
Inger Brandsma et al.
Toxicological sciences : an official journal of the Society of Toxicology, 177(1), 202-213 (2020-07-04)
Understanding the mode-of-action (MOA) of genotoxic compounds and differentiating between direct DNA interaction and indirect genotoxicity is crucial for their reliable safety assessment. ToxTracker is a stem cell-based reporter assay that detects activation of various cellular responses that are associated
Farid Gizatullin et al.
Cancer research, 66(15), 7668-7677 (2006-08-04)
VX-680 is a potent inhibitor of Aurora kinases that induces the accumulation of cells with > or =4N DNA content, followed by cell death. Here, we define the role of p53 and p21(Waf1/Cip1) in cell cycle perturbations following exposure to
Matthew A Young et al.
Cancer research, 66(2), 1007-1014 (2006-01-21)
We present a high-resolution (2.0 A) crystal structure of the catalytic domain of a mutant form of the Abl tyrosine kinase (H396P; Abl-1a numbering) that is resistant to the Abl inhibitor imatinib. The structure is determined in complex with the
Derek Lessing et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(50), 14366-14371 (2017-02-10)
X-chromosome inactivation is a mechanism of dosage compensation in which one of the two X chromosomes in female mammals is transcriptionally silenced. Once established, silencing of the inactive X (Xi) is robust and difficult to reverse pharmacologically. However, the Xi

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service