Skip to Content
Merck
All Photos(1)

Key Documents

SML2157

Sigma-Aldrich

Homoplantaginin

≥98% (HPLC)

Synonym(s):

4′,5,7-Trihydroxy 6-methoxyflavone 7-glucoside, 6-Methoxyapigenin 7-O-β-D-glucoside, 6-Methoxyapigenin 7-O-glucoside, 6-Methylscutellarein 7-glucoside, 7-(β-D-Glucopyranosyloxy)-5-hydroxy-2-(4-hydroxyphenyl)-6-methoxy-4H-1-benzopyran-4-one, Dinatin 7-glucoside, Hispidulin 7-β-glucoside, Hispiduloside, Salvitin 7-glucoside, Scutellarein 6-methyl ether 7-glucoside

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$178.00
25 MG
$569.00

$178.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$178.00
25 MG
$569.00

About This Item

Empirical Formula (Hill Notation):
C22H22O11
CAS Number:
Molecular Weight:
462.40
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$178.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)Oc2cc3[o]c(c[c](c3c(c2OC)O)=O)c4ccc(cc4)O

InChI

1S/C22H22O11/c1-30-21-14(32-22-20(29)19(28)17(26)15(8-23)33-22)7-13-16(18(21)27)11(25)6-12(31-13)9-2-4-10(24)5-3-9/h2-7,15,17,19-20,22-24,26-29H,8H2,1H3/t15-,17-,19+,20-,22-/m1/s1

InChI key

GCLAFEGUXXHIFT-IWLDQSELSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Homoplantaginin is and anti-inflammatory and antioxidant flavonoid isolated from medical herb Salvia plebeian (Sage weed) that improves hepatic injury, hyperlipidemia, and hyperglycemia. Homoplantaginin ameliorates endothelial insulin resistance by inhibiting inflammation and modulating cell signalling via the IKKβ/IRS-1/pAkt/peNOS pathway. Homoplantaginin protects endothelial cells from Palmitic acid -induced inflammation by suppression of TLR4 expression, and NLRP3 inflammasome activation.
anti-inflammatory and antioxidant

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshiaki Amakura et al.
Molecules (Basel, Switzerland), 19(4), 4956-4966 (2014-04-22)
Many recent studies have suggested that activation of the aryl hydrocarbon receptor (AhR) reduces immune responses, thus suppressing allergies and autoimmune diseases. In our continuing study on natural AhR agonists in foods, we examined the influence of 37 health food
Feihua Wu et al.
Biological & pharmaceutical bulletin, 35(7), 1171-1177 (2012-07-14)
Recent data have indicated that inflammation plays an important role in the development of insulin resistance. The present study aims at examining the activity of homoplantaginin, a flavonoid from a traditional Chinese medicine Salvia plebeia R. BR., on palmitic acid
Baiqiu He et al.
Journal of cardiovascular pharmacology, 67(1), 93-101 (2015-09-12)
Palmitic acid (PA)-induced vascular endothelial inflammation plays a pivotal role in the occurrence and development of vascular diseases. The present study was conducted to examine the effect of homoplantaginin, a main flavonoid from a traditional Chinese medicine Salvia plebeia R.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service