Skip to Content
Merck
All Photos(1)

Key Documents

SML2150

Sigma-Aldrich

AZ6102

≥98% (HPLC)

Synonym(s):

rel-2-[4-[6-[(3R,5S)-3,5-Dimethyl-1-piperazinyl]-4-methyl-3-pyridinyl]phenyl]-3,7-dihydro-7-methyl-4H-pyrrolo[2,3-d]pyrimidin-4-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$106.00
25 MG
$430.00

$106.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$106.00
25 MG
$430.00

About This Item

Empirical Formula (Hill Notation):
C25H28N6O
CAS Number:
Molecular Weight:
428.53
UNSPSC Code:
12352200
NACRES:
NA.77

$106.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CN1C=CC2=C1N=C(C3=CC=C(C4=CN=C(N5C[C@H](C)N[C@H](C)C5)C=C4C)C=C3)NC2=O

Biochem/physiol Actions

AZ6102 is a potent and selective Tankyrase 1/2 (TAMKS1/2) inhibitor that has 100-fold selectivity against other PARP family enzymes. AZ6102 is moderately orally bioavailable in rat and mice.
bioavailable, potent and selective Tankyrase 1/2 (TAMKS1/2) inhibitor

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeffrey W Johannes et al.
ACS medicinal chemistry letters, 6(3), 254-259 (2015-03-31)
The canonical Wnt pathway plays an important role in embryonic development, adult tissue homeostasis, and cancer. Germline mutations of several Wnt pathway components, such as Axin, APC, and ß-catenin, can lead to oncogenesis. Inhibition of the poly(ADP-ribose) polymerase (PARP) catalytic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service