Skip to Content
Merck
All Photos(1)

Key Documents

SML2111

Sigma-Aldrich

GSK8814

≥98% (HPLC)

Synonym(s):

8-((3R,4R,5S)-3-((4,4-Difluorocyclohexyl)methoxy)-5-methoxypiperidin-4-ylamino)-3-methyl-5-(5-methylpyridin-3-yl)-1,7-naphthyridin-2(1H)-one, 8-[[(3R,4R,5S)-3-[(4,4-Difluorocyclohexyl)methoxy]-5-methoxy-4-piperidinyl]amino]-3-methyl-5-(5-methyl-3-pyridinyl)-1,7-naphthyridin-2(1H)-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$278.00
25 MG
$1,120.00

$278.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$278.00
25 MG
$1,120.00

About This Item

Empirical Formula (Hill Notation):
C28H35F2N5O3
CAS Number:
Molecular Weight:
527.61
UNSPSC Code:
51111800
NACRES:
NA.77

$278.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

colorless to very dark brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(N1)C(C)=CC2=C1C(N[C@H]3[C@H](OCC4CCC(F)(F)CC4)CNC[C@@H]3OC)=NC=C2C5=CN=CC(C)=C5

Biochem/physiol Actions

Aqueous soluble inhibitor that targets ATAD2 & ATAD2B bromodomain with high selectivity and submicromolar affinity.
GSK8814 is an aqueous soluble (>439 μM) inhibitor that targets ATAD2 & ATAD2B bromodomain (BD) with submicromolar affinity (pKd = 8.1 for ATAD2 by ITC) and high selectivity (pIC50 = 7.3/ATAD2 BD & 7.7/ATAD2B BD in competitive ligand binding assays by TR-FRET; pIC50 ≤4.5 when using BD1/BD2 of BRD2-4 or BRDT). When tested using intact cells, GSK8814 effectively disrupts histone H3.3 interaction with ATAD2 BD construct (IC50 = 2.7 μM), but not full-length ATAD2. GSK8814, but not its less active diastereomer GSK8815, is shown to suppress LNCaP colony formation in a soft agar assay, albeit with a high effective concentration (by 56% at 20 μM).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul Bamborough et al.
Angewandte Chemie (International ed. in English), 55(38), 11382-11386 (2016-08-18)
ATAD2 is a cancer-associated protein whose bromodomain has been described as among the least druggable of that target class. Starting from a potent lead, permeability and selectivity were improved through a dual approach: 1) using CF2 as a sulfone bio-isostere

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service