Skip to Content
Merck
All Photos(1)

Key Documents

SML2075

Sigma-Aldrich

Gimeracil

≥98% (HPLC)

Synonym(s):

5-Chloropyridine-2,4-diol, 5-chloro-4-hydroxypyridin-2(1H)-one, CDHP, Gimestat

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$74.10
25 MG
$181.00

$74.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$74.10
25 MG
$181.00

About This Item

Empirical Formula (Hill Notation):
C5H4ClNO2
CAS Number:
Molecular Weight:
145.54
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$74.10


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

Clc1c[nH][c](cc1O)=O

InChI

1S/C5H4ClNO2/c6-3-2-7-5(9)1-4(3)8/h1-2H,(H2,7,8,9)

InChI key

ZPLQIPFOCGIIHV-UHFFFAOYSA-N

Biochem/physiol Actions

Gimeracil (5-chloro-2, 4-dihydroxypyridine) is an orally active inhibitor against dihydropyrimidine dehydrogenase (DPYD), which degrades pyrimidine and the chemotherapeutic drug 5-fluorouracil (5FU). Gimeracil is shown to exhibit cancer-radiosensitizing efficacy in cultures in vitro (1 mM) and in mice in vivo (2.5-25 mg/kg, p.o.) in a DPYD-dependent manner via partial suppression of DNA repair by homologous recombination (HR).
Orally active dihydropyrimidine dehydrogenase (DPYD) inhibitor with cancer-radiosensitizing efficacy in vitro and in vivo.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masakazu Fukushima et al.
Oncology reports, 24(5), 1307-1313 (2010-09-30)
Chemoradiotherapy is a useful treatment strategy in patients with locally advanced cancers. In particular, combination of 5-fluorouracil (5-FU) with X-ray irradiation is effective for the treatment of some types of gastrointestinal cancers. We investigated the antitumor effects of combination treatment
Keiji Koda et al.
Anticancer research, 36(10), 5325-5331 (2016-11-01)
It has not been elucidated whether the clinical efficacy of oral fluoropyrimidines for adjuvant chemotherapy of colorectal cancer varies with tumor biological characteristics. A multicenter randomized trial was performed comparing oral tegafur/gimeracil/oteracil (S-1) and uracil-tegafur/ leucovorin (UFT/LV) as adjuvant therapy
Ryohei Nishiguchi et al.
BMJ case reports, 2016 (2016-12-10)
The most common histological classification of bile duct cancer is adenocarcinoma and squamous cell carcinoma (SCC) is relatively rare. We report a case of a 78-year-old man with SCC of the extrahepatic bile duct associated with metachronous para-aortic lymph node
T Wang et al.
Cancer radiotherapie : journal de la Societe francaise de radiotherapie oncologique, 20(3), 181-186 (2016-04-14)
This study evaluated the efficacy and safety of the combination drug tegafur, gimeracil, and oteracil potassium (S-1) concurrent with 3-dimensional conformal radiotherapy for newly diagnosed squamous cell carcinoma of the lung in elderly patients. Patients with pathologically or cytologically newly
Koh-Ichi Sakata et al.
Cancer science, 102(9), 1712-1716 (2011-06-15)
Gimeracil (5-chloro-2, 4-dihydroxypyridine) is an inhibitor of dihydropyrimidine dehydrogenase (DPYD), which degrades pyrimidine including 5-fluorouracil in the blood. Gimeracil was originally added to an oral fluoropyrimidine derivative S-1 to yield prolonged 5-fluorouracil concentrations in serum and tumor tissues. We have

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service