Skip to Content
Merck
All Photos(1)

Key Documents

SML2047

Sigma-Aldrich

BPAM344

≥98% (HPLC)

Synonym(s):

BPAM344, 4-Cyclopropyl-3,4-dihydro-7-fluoro-2H-1,2,4-benzothiadiazine 1,1-dioxide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$160.00

$160.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$160.00

About This Item

Empirical Formula (Hill Notation):
C10H11FN2O2S
CAS Number:
Molecular Weight:
242.27
UNSPSC Code:
12352200
NACRES:
NA.77

$160.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

storage temp.

2-8°C

SMILES string

O=S1(C2=CC(F)=CC=C2N(C3CC3)CN1)=O

InChI

1S/C10H11FN2O2S/c11-7-1-4-9-10(5-7)16(14,15)12-6-13(9)8-2-3-8/h1,4-5,8,12H,2-3,6H2

InChI key

FLTMTBPCYAZIKM-UHFFFAOYSA-N

Biochem/physiol Actions

BPAM344 is a potent positive allosteric modulator (PAM) on the three KAR subunits GluK1b, GluK2a, and GluK3a. BPAM344 potentiates glutamate-evoked currents of GluK2a 21-fold, and noticeably decreases desensitization kinetics (from 5.5 to 775 ms).
Potent positive allosteric modulator (PAM) on the three KAR subunits GluK1b, GluK2a, and GluK3a

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian Krintel et al.
Biophysical journal, 110(11), 2397-2406 (2016-06-09)
The 1,2,4-benzothiadiazine 1,1-dioxide type of positive allosteric modulators of the ionotropic glutamate receptor A2 (GluA2) are promising lead compounds for the treatment of cognitive disorders, e.g., Alzheimer's disease. The modulators bind in a cleft formed by the interface of two
Ann-Beth Nørholm et al.
Journal of medicinal chemistry, 56(21), 8736-8745 (2013-10-18)
Positive allosteric modulators of ionotropic glutamate receptors are potential compounds for treatment of cognitive disorders, e.g., Alzheimer's disease. The modulators bind within the dimer interface of the ligand-binding domain (LBD) and stabilize the agonist-bound conformation, thereby slowing receptor desensitization and/or
Anja Probst Larsen et al.
Molecular pharmacology, 91(6), 576-585 (2017-04-01)
Kainate receptors (KARs) consist of a class of ionotropic glutamate receptors, which exert diverse pre- and postsynaptic functions through complex signaling regulating the activity of neural circuits. Whereas numerous small-molecule positive allosteric modulators of the ligand-binding domain of (

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service