Skip to Content
Merck
All Photos(1)

Key Documents

SML2044

Sigma-Aldrich

DMAT

≥98% (HPLC)

Synonym(s):

CK2 Inhibitor II, 2-Dimethylamino-4,5,6,7-tetrabromo-1H-benzimidazole, 4,5,6,7-Tetrabromo-N,N-dimethyl-1H-benzimidazol-2-amine, 4,5,6,7-Tetrabromo-N,N-dimethyl-1H-benzo[d]imidazol-2-amine, DMAT

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$132.00
25 MG
$428.00

$132.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$132.00
25 MG
$428.00

About This Item

Empirical Formula (Hill Notation):
C9H7Br4N3
CAS Number:
Molecular Weight:
476.79
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$132.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

shipped in

wet ice

storage temp.

−20°C

SMILES string

CN(C)c1nc2c(Br)c(Br)c(Br)c(Br)c2[nH]1

InChI

1S/C9H7Br4N3/c1-16(2)9-14-7-5(12)3(10)4(11)6(13)8(7)15-9/h1-2H3,(H,14,15)

InChI key

SLPJGDQJLTYWCI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

DMAT has been used as a casein kinase 2 (CK2) inhibitor:
  • to study its effects on parasite multiplication in a [3H]-hypoxanthine incorporation assay[1]
  • to study its effects on the intracellular response in prostate cancer cells[2]
  • to study its effects on cell division cycle 25C (cdc25C) phosphatase levels in prostate cancer cells[3]

Biochem/physiol Actions

Casein kinase II inhibitor II (Ck2 Inhibitor II) is a ATP competitive inhibitor of casein kinase II.
Casein kinase II inhibitor II (Ck2 Inhibitor II) is a high affinity ATP-competitive inhibitor of casein kinase II. Ck2 Inhibitor II inhibits CK2 in rat liver with 1,300-fold greater selectivity for CK2 than for CK1 (IC50 = 140 nM and >200 μM respectively). Ki = 40 nM; In Jurkat human T-cell leukemia cells, Ck2 Inhibitor II does not display side effects on mitochondria polarization at concentrations up to 10 μM; Ck2 Inhibitor II is useful for in vivo studies due to cell permeability and high efficacy in cultured cells and has implications in research and treatment of neoplasia and infective diseases, in which CK2 plays a role and is highly active constitutively.
DMAT is an ATP-competitive (Ki = 40 nM) casein kinase 2 (CK2) inhibitor (IC50 = 140 nM; rat liver CK2, [ATP] = 20 nM) with greatly improved potency and selectivity than its structure analog TBB. With the exception of DYRK1a (IC50 = 120 nM), DMAT is reported to exhibit no CK1 inhibitiory potency up to 200 μM (50% inhibition by 29 μM TBB) and little or no activity against a panel of 32 other protein kinases, nor PI3K α and γ. When tested in Jurkat cultures, DMAT is shown to be a superior apoptosis inducer than TBB (DC50 = 2.7 μM/DMAT vs. 17 μM/TBB).

Caution

Air sensitive

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander Ulges et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(36), 10145-10150 (2016-08-25)
T helper 17 (TH17) cells represent a discrete TH cell subset instrumental in the immune response to extracellular bacteria and fungi. However, TH17 cells are considered to be detrimentally involved in autoimmune diseases like multiple sclerosis (MS). In contrast to
Carolin C Schneider et al.
Molecular and cellular biochemistry, 356(1-2), 177-184 (2011-07-14)
Protein kinase CK2 is implicated in the regulation of the cell cycle. In addition to a variety of functions, CK2 has anti-apoptotic properties. So far the role of CK2 linking both pathways in the cell is not clear. Some years
Mario A Pagano et al.
Journal of medicinal chemistry, 47(25), 6239-6247 (2004-11-30)
Casein kinase 2 (CK2) is a ubiquitous, essential, and highly pleiotropic protein kinase whose abnormally high constitutive activity is suspected to underlie its pathogenic potential in neoplasia and infective diseases. Thus, CK2 inhibitors designed to dissect the signaling pathways affected
Mario A Pagano et al.
Biochemical and biophysical research communications, 321(4), 1040-1044 (2004-09-11)
Protein kinase CK2 is a highly pleiotropic enzyme whose high constitutive activity is suspected to be instrumental to the enhancement of the tumour phenotype and to the propagation of infectious diseases. Here we describe a novel compound, 2-dimethylamino-4,5,6,7-tetrabromo-1H-benzimidazole (DMAT), which
Claudia Götz et al.
Biochimica et biophysica acta, 1820(7), 970-977 (2012-03-06)
Abnormally high activity of protein kinase CK2 is linked to various diseases including cancer. Therefore, the inhibition of CK2 is a promising therapeutic strategy to fight this disease. We screened a library of synthetic molecules concerning their capacity to inhibit

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service