Skip to Content
Merck
All Photos(1)

Key Documents

SML2043

Sigma-Aldrich

CID 1375606

≥98% (HPLC)

Synonym(s):

2,4-Dichloro-N-[4-[(phenylamino)carbonyl]phenyl]-benzamide, N-[4-(Anilinocarbonyl)phenyl]-2,4-dichlorobenzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H14Cl2N2O2
CAS Number:
Molecular Weight:
385.24
UNSPSC Code:
12352202
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC1=CC=C(C(NC2=CC=C(C(NC3=CC=CC=C3)=O)C=C2)=O)C(Cl)=C1

Biochem/physiol Actions

CID 1375606 (N-[4-(Anilinocarbonyl)phenyl]-2,4-dichlorobenzamide) is a specific and potent surrogate agonist of GPR27 (SREB1) that induces coupling of the receptor to β-arrestin 2 but not to G proteins.
Specific and potent surrogate agonist of GPR27 (SREB1).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service