Skip to Content
Merck
All Photos(1)

Key Documents

SML2021

Sigma-Aldrich

MIND4-17

≥98% (HPLC)

Synonym(s):

5-Nitro-2-{[5-(phenoxymethyl)-4-phenyl-4H-1,2,4-triazol-3-yl]thio}pyridine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$182.00
25 MG
$484.00

$182.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$182.00
25 MG
$484.00

About This Item

Empirical Formula (Hill Notation):
C20H15N5O3S
CAS Number:
Molecular Weight:
405.43
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$182.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to brown

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O=[N+]([O-])C(C=C1)=CN=C1SC2=NN=C(COC3=CC=CC=C3)N2C4=CC=CC=C4

InChI

1S/C20H15N5O3S/c26-25(27)16-11-12-19(21-13-16)29-20-23-22-18(14-28-17-9-5-2-6-10-17)24(20)15-7-3-1-4-8-15/h1-13H,14H2

InChI key

OZUBDKIROJPQGE-UHFFFAOYSA-N

Biochem/physiol Actions

KEAP1 covalent modifier with therapeutic potential for Huntington′s disease by promoting NRF2 activation.
MIND4-17 is a potent NRF2 (nuclear factor erythroid 2-related factor 2) activator that covalently modifies a critical stress-sensor cysteine (C151) in the BTB domain of the E3 ligase substrate adaptor protein Kelch-like ECH-associated protein 1 (KEAP1), the primary negative regulator of NRF2. MIND4-17 induces NRF2 activation responses in neuronal and non-neuronal cultures (effective conc. 0.1-10 μM) of human, mouse, and rat origins. In addition, MIND4-17 effectively reduces endotoxin-induced IL-6 release from WT as well as YAC128 HD mutant mice-derived primary microglia and astrocytes, however, NRF2 induction by MIND4-17 is shown to be compromised in human Huntington′s diseased (HD) relative to non-disesed neural stem cells due to suppressive influence of the expanded CAG repeat mutation on pathway activation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luisa Quinti et al.
Cell chemical biology, 23(7), 849-861 (2016-07-19)
There are currently no disease-modifying therapies for the neurodegenerative disorder Huntington's disease (HD). This study identified novel thiazole-containing inhibitors of the deacetylase sirtuin-2 (SIRT2) with neuroprotective activity in ex vivo brain slice and Drosophila models of HD. A systems biology approach
Luisa Quinti et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(23), E4676-E4685 (2017-05-24)
The activity of the transcription factor nuclear factor-erythroid 2 p45-derived factor 2 (NRF2) is orchestrated and amplified through enhanced transcription of antioxidant and antiinflammatory target genes. The present study has characterized a triazole-containing inducer of NRF2 and elucidated the mechanism

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service