Skip to Content
Merck
All Photos(1)

Key Documents

SML1974

Sigma-Aldrich

CBLC000 trifluoroacetate

≥95% (HPLC)

Synonym(s):

9H-Carbazole-9-ethanamine, N,N-diethyl-3,6-dinitro- trifluoroacetate, Curaxin CBLC000 trifluoroacetate, N,N-Diethyl-3,6-dinitro-9H-carbazole-9-ethanamine trifluoroacetate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$120.00

$120.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$120.00

About This Item

Empirical Formula (Hill Notation):
C18H20N4O4 · C2HF3O2
CAS Number:
Molecular Weight:
470.40
UNSPSC Code:
12352200
NACRES:
NA.77

$120.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CCN(CC)CCN1C2=C(C=C([N+]([O-])=O)C=C2)C3=C1C=CC([N+]([O-])=O)=C3.FC(F)(C(O)=O)F

InChI

1S/C18H20N4O4.C2HF3O2/c1-3-19(4-2)9-10-20-17-7-5-13(21(23)24)11-15(17)16-12-14(22(25)26)6-8-18(16)20;3-2(4,5)1(6)7/h5-8,11-12H,3-4,9-10H2,1-2H3;(H,6,7)

InChI key

VFGJIBCWHYFSHE-UHFFFAOYSA-N

Biochem/physiol Actions

CBLC000 is a potent inhibitor of FACT (facilitates chromatin transcription) complex that activates p53 and suppresses NF-kB without genotoxicity. CBLC000 induces chromatin trapping of FACT both in vitro and in vivo. CBLC000 exhibits brad anticancer activity. CBLC000 analog clinical candidate CBL0137 decreases self-renewal of GBM stem cells (GSC).
Potent inhibitor of FACT complex that activates p53 and suppresses NF-kB without genotoxicity

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander V Gasparian et al.
Science translational medicine, 3(95), 95ra74-95ra74 (2011-08-13)
Effective eradication of cancer requires treatment directed against multiple targets. The p53 and nuclear factor κB (NF-κB) pathways are dysregulated in nearly all tumors, making them attractive targets for therapeutic activation and inhibition, respectively. We have isolated and structurally optimized
Josephine Kam Tai Dermawan et al.
Cancer research, 76(8), 2432-2442 (2016-02-28)
The nearly universal recurrence of glioblastoma (GBM) is driven in part by a treatment-resistant subpopulation of GBM stem cells (GSC). To identify improved therapeutic possibilities, we combined the EGFR/HER2 inhibitor lapatinib with a novel small molecule, CBL0137, which inhibits FACT

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service