Skip to Content
Merck
All Photos(1)

Key Documents

SML1971

Sigma-Aldrich

LLY-284

≥98% (HPLC)

Synonym(s):

(2R,3R,4S,5R)-2-(4-Amino-7H-pyrrolo[2,3-d]pyrimidin-7-yl)-5-((S)-hydroxy(phenyl)methyl)tetrahydrofuran-3,4-diol, (S)-5′-phenyl-7-deazaadenosine; 6-amino-9-[(S)-5′-phenyl(ribofuranosyl)]-7-deazapurine, LLY 284, LLY284

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$349.00
25 MG
$1,360.00

$349.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$349.00
25 MG
$1,360.00

About This Item

Empirical Formula (Hill Notation):
C17H18N4O4
CAS Number:
Molecular Weight:
342.35
UNSPSC Code:
12352200
NACRES:
NA.77

$349.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

storage temp.

2-8°C

SMILES string

O[C@@H]1[C@@H]([C@@H](O)C2=CC=CC=C2)O[C@@H](N3C(N=CN=C4N)=C4C=C3)[C@@H]1O

InChI key

WWOOWAHTEXIWBO-CNUKPYSBSA-N

Biochem/physiol Actions

LLY-284 is the less potent diastereomer of the potent SAM-competitive PRMT5 inhibitor LLY-283 (IC50 = 20 nM against PRMT5-catalyzed methylation of an H4R3-derived peptide substrate) and an ideal control compound for cellular studies. For characterization details of the active probe, LLY-283, please visit the LLY-283 probe summary on the Structural Genomics Consortium (SGC) website.

LLY-283, the active probe, is available from Sigma. To learn more about and purchase LLY-283, click here.

To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc
Less potent diastereomer and control compound for the potent and selective SAM-competitive PRMT5 inhibitor LLY-283.

Features and Benefits

LLY-284 is a negative control for LLY-283, an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service