Skip to Content
Merck
All Photos(1)

Documents

SML1968

Sigma-Aldrich

Phentermine hydrochloride

≥98% (HPLC)

Synonym(s):

α,α-Dimethylphenethylamine hydrochloride, α-Benzylisopropylamine hydrochloride, 1,1-Dimethyl-2-phenylethylamine hydrochloride, 2-Methyl-1-phenylpropan-2-amine hydrochloride, 2-Phenyl-tert-butylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15N · HCl
CAS Number:
Molecular Weight:
185.69
EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Cl.CC(C)(N)Cc1ccccc1

InChI

1S/C10H15N.ClH/c1-10(2,11)8-9-6-4-3-5-7-9;/h3-7H,8,11H2,1-2H3;1H

InChI key

NCAIGTHBQTXTLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Phentermine is a sympathomimetic that releases norepinephrine and epinephrine in CNS and periphery reducing hunger. Similarly to other amphetamines, phentermine potently inhibits DA and 5-HT uptake.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Candida J Rebello et al.
Advances in therapy, 33(11), 1853-1866 (2016-10-28)
The homeostatic controls over eating are inextricably linked to the reward aspects of eating. The result is an integrated response that coordinates the internal milieu with the prevailing environment. Thus, appetite, which reflects a complex interaction among the external environment
Mark J Ferris et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 37(7), 1708-1716 (2012-03-08)
The dopamine transporter (DAT) is the primary site of action for psychostimulant drugs such as cocaine, methylphenidate, and amphetamine. Our previous work demonstrated a reduced ability of cocaine to inhibit the DAT following high-dose cocaine self-administration (SA), corresponding to a
B Kalyanasundar et al.
Journal of neurophysiology, 114(1), 585-607 (2015-05-15)
Obesity is a worldwide health problem that has reached epidemic proportions. To ameliorate this problem, one approach is the use of appetite suppressants. These compounds are frequently amphetamine congeners such as diethylpropion (DEP), phentermine (PHEN), and bupropion (BUP), whose effects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service