Skip to Content
Merck
All Photos(1)

Key Documents

SML1960

Sigma-Aldrich

GSK583

≥98% (HPLC)

Synonym(s):

GSK583, 6-(tert-Butylsulfonyl)-N-(5-fluoro-1H-indazol-3-yl)quinolin-4-amine, GSK′583, GSK-2616583A

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$189.00
25 MG
$605.00

$189.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$189.00
25 MG
$605.00

About This Item

Empirical Formula (Hill Notation):
C20H19FN4O2S
CAS Number:
Molecular Weight:
398.45
UNSPSC Code:
51111800
NACRES:
NA.77

$189.00


Estimated to ship on02 June 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated
protect from light

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

Fc1cc2c([nH]nc2Nc3c4c(ncc3)ccc(c4)[S](=O)(=O)C(C)(C)C)cc1

InChI key

XLOGLWKOHPIJLV-UHFFFAOYSA-N

Biochem/physiol Actions

An ATP-competitive RIP2 (RIPK2) inhibitor with in vitro and in vivo efficacy against NOD1- and NOD2-dependent pro-inflammatory cytokines release.
GSK583 is a cell-permeable indazolylquinolinamine that acts as an ATP-competitive (IC50 = 2 & 5 nM using rat & human RIPK2, respectively) inhibitor against RIPK2 kinase activity (66.6% inhibition at 1 μM; [ATP] = 10 μM), exhibiting much reduced or no inhibitory potency against a panel of 209 other kinases <34% inhibition at 1 μM; [ATP] = 10 ?M). GSK583 selectively inhibits against NOD1- and NOD2-, but not TLR2-, TLR4-, TLR7-. IL-1R-, or TNFR-dependent pro-inflammatory cytokines release (>95% inhibition vs. <40% inhibition at 1 ?M using primary human monocytes stimulated with respective ligands). GSK583 effectively inhibits TNF? and IL-6 production from Crohn′s disease (CD) and ulcerative colitis (UC) patients intestinal explant cultures ex vivo (IC50 = 200 nM) and is efficacious against NOD1 ligand FK156- and NOD2 ligand MDP-induced inflammatory responses in mice and rats (10 mg/kg p.o.) in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service