Skip to Content
Merck
All Photos(1)

Key Documents

SML1945

Sigma-Aldrich

IMB-808

≥98% (HPLC)

Synonym(s):

2,3-Dihydro-N-methyl-N-[2-oxo-2-[(2,3,4-trifluorophenyl)amino]ethyl]-1,4-benzodioxin-6-carboxamide, N-Methyl-N-(2-oxo-2-((2,3,4-trifluorophenyl)amino)ethyl)-3,4-dihydro-2H-benzo[b][1,4] dioxin-7-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$120.00

$120.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$120.00

About This Item

Empirical Formula (Hill Notation):
C18H15F3N2O4
CAS Number:
Molecular Weight:
380.32
UNSPSC Code:
12352200
NACRES:
NA.77

$120.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC1=C(NC(CN(C)C(C2=CC(OCCO3)=C3C=C2)=O)=O)C=CC(F)=C1F

InChI

1S/C18H15F3N2O4/c1-23(9-15(24)22-12-4-3-11(19)16(20)17(12)21)18(25)10-2-5-13-14(8-10)27-7-6-26-13/h2-5,8H,6-7,9H2,1H3,(H,22,24)

InChI key

XITZPIXICJOKTF-UHFFFAOYSA-N

Biochem/physiol Actions

IMB-808 is a potent Liver X receptor α and β (LXRα/β) partial dual agonist that promotes expression of genes related to reverse cholesterol transport (including ABCA1 and ABCG1), and associated with cholesterol metabolism pathway in multiple cell lines (including ApoE). Also IMB-808 promotes cholesterol efflux from macrophages and reduced the accumulated lipids of foam cells.
potent Liver X receptor α and β (LXRα/β) partial dual agonist

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ni Li et al.
Molecular pharmacology, 91(4), 264-276 (2017-01-15)
Activation of liver X receptor (LXR) is associated with cholesterol metabolism and anti-inflammatory processes, which makes it beneficial to antiatherosclerosis therapy. Nevertheless, existing agonists that target LXR, for example TO901317, are related to unwanted side effects. In the present study

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service