Skip to Content
Merck
All Photos(1)

Key Documents

SML1944

Sigma-Aldrich

Bz-423

≥98% (HPLC)

Synonym(s):

7-Chloro-1,3-dihydro-5-(4-hydroxyphenyl)-1-methyl-3-(2-naphthalenylmethyl)-2H-1,4-benzodiazepin-2-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$178.00
25 MG
$569.00

$178.00


Available to ship on07 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 MG
$178.00
25 MG
$569.00

About This Item

Empirical Formula (Hill Notation):
C27H21ClN2O2
CAS Number:
Molecular Weight:
440.92
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$178.00


Available to ship on07 April 2025Details


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC1=CC=C2C(C(C3=CC=C(O)C=C3)=NC(CC4=CC=C(C=CC=C5)C5=C4)C(N2C)=O)=C1

InChI

1S/C27H21ClN2O2/c1-30-25-13-10-21(28)16-23(25)26(19-8-11-22(31)12-9-19)29-24(27(30)32)15-17-6-7-18-4-2-3-5-20(18)14-17/h2-14,16,24,29H,15H2,1H3

InChI key

IJEUIRDNQUQMFU-UHFFFAOYSA-N

Application

Bz-423 has been used as a mitochondrial inner membrane pore (MIMP) reagent to study its effect on oxygen consumption rate (OCR) and ATP synthesis in HepG2 cells.[1]

Biochem/physiol Actions

Bz-423 is an inhibitor of mitochondrial F1F0 ATP synthase. It binds the oligomycin sensitivity conferring protein (OSCP) component of mitochondrial F1F0-ATPase, resulting in superoxide generation and apoptosis. BZ-423 was shown to block multidrug-resistant melanoma cells. In another study, Bz-423 selectively increased superoxide and induced the apoptosis of alloreactive T cells, which arrested established Graft versus Host Disease.
Bz-423 stimulates mitochondrial inner membrane pore (MIMP) opening in isolated mitochondria.[1]
Inhibitor of mitochondrial F1F0-ATP synthase

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anthony J Bilotta et al.
Cellular and molecular gastroenterology and hepatology, 11(4), 1023-1044 (2020-11-26)
Gut bacteria-derived short-chain fatty acids (SCFAs) play crucial roles in the maintenance of intestinal homeostasis. However, how SCFAs regulate epithelial turnover and tissue repair remain incompletely understood. In this study, we investigated how the SCFA propionate regulates cell migration to
Abby Hearne et al.
Toxicology in vitro : an international journal published in association with BIBRA, 67, 104907-104907 (2020-06-06)
Oligomycin is a classical mitochondrial reagent that binds to the proton channel on the Fo component of ATP synthase. As a result, oligomycin blocks mitochondrial ATP synthesis, proton translocation, and O2 uptake. Here we show that oligomycin induces proton uncoupling

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service