Skip to Content
Merck
All Photos(1)

Key Documents

SML1897

Sigma-Aldrich

NSC73306

≥98% (HPLC)

Synonym(s):

2-(1,2-Dihydro-2-oxo-3H-indol-3-ylidene)-N-(4-methoxyphenyl)hydrazinecarbothioamide, NSC 671054, NSC 73304

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$136.00

$136.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$136.00

About This Item

Empirical Formula (Hill Notation):
C16H14N4O2S
CAS Number:
Molecular Weight:
326.37
UNSPSC Code:
12352202
NACRES:
NA.77

$136.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, yellow to light brown

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

S=C(N\N=C2\c3c(c(cc(c3)Cl)Cl)NC\2=O)Nc1ccc(cc1)OC

InChI

1S/C16H12Cl2N4O2S/c1-24-10-4-2-9(3-5-10)19-16(25)22-21-14-11-6-8(17)7-12(18)13(11)20-15(14)23/h2-7H,1H3,(H2,19,22,25)(H,20,21,23)

InChI key

HPXWWBMVIMFLLU-UHFFFAOYSA-N

Biochem/physiol Actions

NSC73306, a thiosemicarbazone, is a cell penetrant, cytotoxic agent that exhibits greater toxicity against cells expressing functional P-gp (P-glycoprotein) than against other cells.
NSC73306, a thiosemicarbazone, is a cell penetrant, cytotoxic agent that exhibits greater toxicity against cells expressing functional P-gp (P-glycoprotein) than against other cells. Irrespective of variations in cell line background, NSC73306 consistently demonstrated a Pgp-potentiated MDR-selective toxicity. It appears that NSC73306 is transported into the cell by copper transporter 1 (CTR1, SLC31A1). It is not P-gp inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph A Ludwig et al.
Cancer research, 66(9), 4808-4815 (2006-05-03)
ATP-binding cassette (ABC) proteins include the best known mediators of resistance to anticancer drugs. In particular, ABCB1 [MDR1/P-glycoprotein (P-gp)] extrudes many types of drugs from cancer cells, thereby conferring resistance to those agents. Attempts to overcome P-gp-mediated drug resistance using
Felipe S Pessoto et al.
Oxidative medicine and cellular longevity, 2015, 394367-394367 (2015-06-16)
A series of thiosemicarbazone (TSC) p-substituted acetophenone derivatives were synthesized and chemically characterized. The p-substituents appended to the phenyl group of the TSC structures were hydrogen, fluor, chlorine, methyl, and nitro, producing compounds named TSC-H, TSC-F, TSC-Cl, TSC-Me, and TSC-NO2
King Leung Fung et al.
Molecular pharmaceutics, 11(8), 2692-2702 (2014-05-08)
Acquired drug resistance in cancer continues to be a challenge in cancer therapy, in part due to overexpression of the drug efflux transporter P-glycoprotein (P-gp, MDR1, ABCB1). NSC73306 is a thiosemicarbazone compound that displays greater toxicity against cells expressing functional

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service