Skip to Content
Merck
All Photos(1)

Key Documents

SML1830

Sigma-Aldrich

JJKK-048

≥98% (HPLC)

Synonym(s):

[4-[Bis(1,3-benzodioxol-5-yl)methyl]-1-piperidinyl]-1H-1,2,4-triazol-1-yl-methanone

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$204.00
25 MG
$831.00

$204.00


Check Cart for Availability


Select a Size

Change View
5 MG
$204.00
25 MG
$831.00

About This Item

Empirical Formula (Hill Notation):
C23H22N4O5
CAS Number:
Molecular Weight:
434.44
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$204.00


Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(N1N=CN=C1)N2CCC(C(C3=CC(OCO4)=C4C=C3)C5=CC=C(OCO6)C6=C5)CC2

InChI

1S/C23H22N4O5/c28-23(27-12-24-11-25-27)26-7-5-15(6-8-26)22(16-1-3-18-20(9-16)31-13-29-18)17-2-4-19-21(10-17)32-14-30-19/h1-4,9-12,15,22H,5-8,13-14H2

InChI key

CLSNATLUIXZPMV-UHFFFAOYSA-N

Biochem/physiol Actions

JJKK-048 is a cell penetrant ultrapotent and highly selective inhibitor of monoacylglycerol lipase (MAGL) that exhibits a low cross-reactivity with other endocannabinoid targets. JJKK-048 is an irreversible inhibitor that binds to the active site S122.
Ultrapotent and highly selective inhibitor of monoacylglycerol lipase (MAGL).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Niina Aaltonen et al.
The Journal of pharmacology and experimental therapeutics, 359(1), 62-72 (2016-07-28)
Monoacylglycerol lipase (MAGL) is a serine hydrolase that acts as a principal degradative enzyme for the endocannabinoid 2-arachidonoylglycerol (2-AG). In addition to terminating the signaling function of 2-AG, MAGL liberates arachidonic acid to be used as a primary source for
Niina Aaltonen et al.
Chemistry & biology, 20(3), 379-390 (2013-03-26)
Monoacylglycerol lipase (MAGL) terminates the signaling function of the endocannabinoid, 2-arachidonoylglycerol (2-AG). During 2-AG hydrolysis, MAGL liberates arachidonic acid, feeding the principal substrate for the neuroinflammatory prostaglandins. In cancer cells, MAGL redirects lipid stores toward protumorigenic signaling lipids. Thus MAGL

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service