Skip to Content
Merck
All Photos(1)

Key Documents

SML1812

Sigma-Aldrich

IMR-1

≥98% (HPLC)

Synonym(s):

2-[2-Methoxy-4-[(4-oxo-2-thioxo-5-thiazolidinylidene)methyl]phenoxy]-acetic acid ethyl ester, Inhibitor of Mastermind Recruitment-1

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$204.00
25 MG
$831.00

$204.00


Estimated to ship on01 June 2025



Select a Size

Change View
5 MG
$204.00
25 MG
$831.00

About This Item

Empirical Formula (Hill Notation):
C15H15NO5S2
CAS Number:
Molecular Weight:
353.41
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$204.00


Estimated to ship on01 June 2025


Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(OCC)COC1=CC=C(C=C2SC(NC2=O)=S)C=C1OC

InChI

1S/C15H15NO5S2/c1-3-20-13(17)8-21-10-5-4-9(6-11(10)19-2)7-12-14(18)16-15(22)23-12/h4-7H,3,8H2,1-2H3,(H,16,18,22)

InChI key

QHPJWPQRZMBKTG-UHFFFAOYSA-N

Application

Inhibitor of mastermind recruitment-1 (IMR-1) has been used as an inhibitor of Notch transcription activation complex to downregulate the expression of Notch and downstream protein in pheochromocytoma (PC -12) cells.[1]

Biochem/physiol Actions

IMR-1 is a cell penetrant and potent inhibitor of Notch transcriptional activation complex that disrupted the recruitment of Mastermind-like 1 (Maml1) to the Notch transcriptional activation complex on chromatin. IMR-1 potently inhibits the growth of Notch-dependent cancer cell lines, and inhibits growth of patient-derived tumor xenografts. IMR-1 is hydrolyzed in vivo to acid metabolite IMR-1A.
IMR-1 is a cell penetrant and potent inhibitor of Notch transcriptional activation complex.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruiting Liu et al.
International journal of clinical and experimental pathology, 11(1), 99-109 (2018-01-01)
Background: Parkinson's disease (PD) is a degenerative disease of central nervous system. 1-Methyl-4-phenylpyridine (MPP+) is a non-selective dopaminergic neurotoxin that induces cell injury similar to PD. This study aimed to explore the protective effects of Radix Ophiopogonis polysaccharide extracts (ROP)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service