Skip to Content
Merck
All Photos(1)

Key Documents

SML1811

Sigma-Aldrich

BIQ

≥98% (HPLC)

Synonym(s):

FG 2216, FG-2216, IOX 3, N-[(1-Chloro-4-hydroxy-3-isoquinolinyl)carbonyl]glycine, YM 311, [[(1-Chloro-4-hydroxyisoquinolin-3-yl)carbonyl]amino]acetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$283.00
25 MG
$1,140.00

$283.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$283.00
25 MG
$1,140.00

About This Item

Empirical Formula (Hill Notation):
C12H9ClN2O4
CAS Number:
Molecular Weight:
280.66
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$283.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

OC1=C(C(NCC(O)=O)=O)N=C(Cl)C2=CC=CC=C21

InChI

1S/C12H9ClN2O4/c13-11-7-4-2-1-3-6(7)10(18)9(15-11)12(19)14-5-8(16)17/h1-4,18H,5H2,(H,14,19)(H,16,17)

InChI key

OUQVKRKGTAUJQA-UHFFFAOYSA-N

Biochem/physiol Actions

BIQ (FG-2216) is an orally available and potent inhibitor of prolyl-hydroxylase (PHD).
BIQ (FG-2216) is an orally available and potent inhibitor of prolyl-hydroxylase (PHD). BIQ induces significant and reversible plasma erythropoietin (EPO) levels in vitro and in hemodialysis patients.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wanja M Bernhardt et al.
Journal of the American Society of Nephrology : JASN, 21(12), 2151-2156 (2010-12-01)
The reasons for inadequate production of erythropoietin (EPO) in patients with ESRD are poorly understood. A better understanding of EPO regulation, namely oxygen-dependent hydroxylation of the hypoxia-inducible transcription factor (HIF), may enable targeted pharmacological intervention. Here, we tested the ability
Matthew M Hsieh et al.
Blood, 110(6), 2140-2147 (2007-06-15)
The hypoxia-inducible factor (HIF) pathway is crucial in mitigating the deleterious effects of oxygen deprivation. HIF-alpha is an essential component of the oxygen-sensing mechanisms and under normoxic conditions is targeted for degradation via hydroxylation by HIF-prolyl hydroxylases. Several HIF-prolyl hydroxylase
Mun Chiang Chan et al.
The Journal of biological chemistry, 291(39), 20661-20673 (2016-08-10)
The hypoxia-inducible factor (HIF) system orchestrates cellular responses to hypoxia in animals. HIF is an α/β-heterodimeric transcription factor that regulates the expression of hundreds of genes in a tissue context-dependent manner. The major hypoxia-sensing component of the HIF system involves

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service