Skip to Content
Merck
All Photos(1)

Key Documents

SML1786

Sigma-Aldrich

Centhaquin

≥98% (HPLC)

Synonym(s):

2-[2-[4-(3-Methylphenyl)-1-piperazinyl]ethyl]quinoline, Centhaquine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$203.00
25 MG
$827.00

$203.00


Estimated to ship on29 April 2025



Select a Size

Change View
5 MG
$203.00
25 MG
$827.00

About This Item

Empirical Formula (Hill Notation):
C22H25N3
CAS Number:
Molecular Weight:
331.45
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$203.00


Estimated to ship on29 April 2025


Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CC1=CC=CC(N(CC2)CCN2CCC3=NC4=CC=CC=C4C=C3)=C1

InChI

1S/C22H25N3/c1-18-5-4-7-21(17-18)25-15-13-24(14-16-25)12-11-20-10-9-19-6-2-3-8-22(19)23-20/h2-10,17H,11-16H2,1H3

InChI key

UJNWGFBJUHIJKK-UHFFFAOYSA-N

Biochem/physiol Actions

Centhaquin is a centrally acting hypotensive agent. Centhaquin is an effective resuscitative agent that improves cardiac output, reduces blood lactate levels, and improves survival time of hemorrhaged rats. Centhaquin exhibits antinociception in the mouse mediated through the opioid, the α2A and α2B adrenoceptors.
Centrally acting hypotensive agent

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service