Skip to Content
Merck
All Photos(1)

Key Documents

SML1781

Sigma-Aldrich

PDD00017273

≥98% (HPLC), powder, poly(ADP-ribose) glycohydrolase inhibitor

Synonym(s):

1-[(1,3-Dimethyl-1H-pyrazol-5-yl)methyl]-1,2,3,4-tetrahydro-N-(1-methylcyclopropyl)-3-[(2-methyl-5-thiazolyl)methyl]-2,4-dioxo-6-quinazolinesulfonamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$229.00
25 MG
$910.00

$229.00


Check Cart for Availability


Select a Size

Change View
5 MG
$229.00
25 MG
$910.00

About This Item

Empirical Formula (Hill Notation):
C23H26N6O4S2
CAS Number:
Molecular Weight:
514.62
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$229.00


Check Cart for Availability

Product Name

PDD00017273, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(NC5(CC5)C)c1cc2c([n]([c]([n]([c]2=O)Cc4[s]c(nc4)C)=O)Cc3[n](nc(c3)C)C)cc1

InChI key

IFWUBRBMMNTBRZ-UHFFFAOYSA-N

Biochem/physiol Actions

PDD00017273 is a potent and selective inhibitor of Poly(ADP-ribose) glycohydrolase (PARG), which catalyses hydrolysis of the O-glycosidic linkages of ADP-ribose polymers, reversing the effects of poly(ADP-ribose) polymerases (PARPs).
PDD00017273 is a potent and selective inhibitor of Poly(ADP-ribose) glycohydrolase (PARG), which catalyses hydrolysis of the O-glycosidic linkages of ADP-ribose polymers, reversing the effects of poly(ADP-ribose) polymerases (PARPs). PARG has been shown to be involved in the repair of single strand DNA breaks. PDD00017273 is selective for PARG, with EC50 = 26 nM, compared to values of 30 μM for PARP1 and ARH3.
PDD00017273 is cell permeable in nature. It is used for specific killing of cells defective in certain homologous recombination (HR) proteins such as breast cancer gene 1/2 (BRCA1/2).[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Radiosensitization with an inhibitor of poly (ADP-ribose) glycohydrolase: A comparison with the PARP1/2/3 inhibitor olaparib.
Gravells P, et al.
DNA Repair, 61, 25-36 (2018)
Dominic I James et al.
ACS chemical biology, 11(11), 3179-3190 (2016-10-01)
The enzyme poly(ADP-ribose) glycohydrolase (PARG) performs a critical role in the repair of DNA single strand breaks (SSBs). However, a detailed understanding of its mechanism of action has been hampered by a lack of credible, cell-active chemical probes. Herein, we
Polly Gravells et al.
DNA repair, 61, 25-36 (2017-11-28)
Upon DNA binding the poly(ADP-ribose) polymerase family of enzymes (PARPs) add multiple ADP-ribose subunits to themselves and other acceptor proteins. Inhibitors of PARPs have become an exciting and real prospect for monotherapy and as sensitizers to ionising radiation (IR). The
Tanay Thakar et al.
Nature communications, 11(1), 2147-2147 (2020-05-03)
Upon genotoxic stress, PCNA ubiquitination allows for replication of damaged DNA by recruiting lesion-bypass DNA polymerases. However, PCNA is also ubiquitinated during normal S-phase progression. By employing 293T and RPE1 cells deficient in PCNA ubiquitination, generated through CRISPR/Cas9 gene editing
Evgeniia Prokhorova et al.
Molecular cell, 81(12), 2640-2655 (2021-05-22)
ARH3/ADPRHL2 and PARG are the primary enzymes reversing ADP-ribosylation in vertebrates, yet their functions in vivo remain unclear. ARH3 is the only hydrolase able to remove serine-linked mono(ADP-ribose) (MAR) but is much less efficient than PARG against poly(ADP-ribose) (PAR) chains in vitro.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service