Skip to Content
Merck
All Photos(1)

Key Documents

SML1778

Sigma-Aldrich

SR0987

≥98% (HPLC)

Synonym(s):

2-Chloro-N-(4-(1,1,1,3,3,3-hexafluoro-2-hydroxy-2-propanyl)phenyl)benzamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$203.00
25 MG
$827.00

$203.00


Check Cart for Availability


Select a Size

Change View
5 MG
$203.00
25 MG
$827.00

About This Item

Empirical Formula (Hill Notation):
C16H10ClF6NO2
CAS Number:
Molecular Weight:
397.70
UNSPSC Code:
12352200
NACRES:
NA.77

$203.00


Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

FC(F)(F)C(O)(C(F)(F)F)c1ccc(cc1)NC(=O)c2c(cccc2)Cl

InChI key

IZVBONMVWLTLNT-UHFFFAOYSA-N

Biochem/physiol Actions

SR0987 is a cell-permeable SR1078 analog with ~6-fold improved agonist potency toward T-cell-specific retinoic acid receptor-related orphan receptor ROR?t (EC50 ~800 nM in cell-based reporter assays) by targeting an allosteric pocket within the RORγt ligand-binding domain (LBD). In addition to upregulating RORγt target gene IL17A expression, SR0987 is shown to simultaneously downregulate surface immune checkpoint protein PD-1 expression by a yet unidentified mechanism among ex vivo differentiated murine TH17 cells, as well as PMA/ionophore-stimulated human Jurkat T cells and murine EL4 T lymphocytes.
SR0987 is a cell-permeable SR1078 analog with ~6-fold improved agonist potency.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service