Skip to Content
Merck
All Photos(1)

Key Documents

SML1753

Sigma-Aldrich

XMD17-109

≥98% (HPLC)

Synonym(s):

11-Cyclopentyl-2-((2-ethoxy-4-(4-(4-methylpiperazin-1-yl)piperidine-1-carbonyl)-phenyl)amino)-5-methyl-5H-benzo[e]pyrimido[5,4-b][1,4]diazepin-6(11H)-one, ERK5-IN-1

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$257.00

$257.00


Check Cart for Availability


Select a Size

Change View
5 MG
$257.00

About This Item

Empirical Formula (Hill Notation):
C36H46N8O3
CAS Number:
Molecular Weight:
638.80
UNSPSC Code:
12352200
NACRES:
NA.77

$257.00


Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

N7(CCN(CC7)C)C1CCN(CC1)C(=O)c2cc(c(cc2)Nc3nc4[n](c6c([c]([n](c4cn3)C)=O)cccc6)C5CCCC5)OCC

InChI

1S/C36H46N8O3/c1-4-47-32-23-25(34(45)43-17-15-26(16-18-43)42-21-19-40(2)20-22-42)13-14-29(32)38-36-37-24-31-33(39-36)44(27-9-5-6-10-27)30-12-8-7-11-28(30)35(46)41(31)3/h7-8,11-14,23-24,26-27H,4-6,9-10,15-22H2,1-3H3,(H,37,38,39)

InChI key

XVBGRTMNFNMINE-UHFFFAOYSA-N

Biochem/physiol Actions

XMD17-109 is a cell-permeable, non-cytotoxic ATP site-targeting pyrimido-diazepinone derivative that acts as a potent inhibitor against ERK5 (MAPK7, BMK1) kinase activity in cell-free assay, and EGF-induced ERK5 autophosphorylation in HeLa cells.
XMD17-109 is a cell-permeable, non-cytotoxic ATP site-targeting pyrimido-diazepinone derivative that acts as potent inhibitor against ERK5 (MAPK7, BMK1) kinase activity in cell-free assay (IC50 = 162 nM; [ATP] = 50 μM) and EGF-induced ERK5 autophosphorylation in HeLa cells (IC50  = 90 nM). XMD17-109 displayed excellent target selectivity on a 442-kinase panel. XMD17-109 effectively inhibits inflammatory stimuli-induced IL-6 and IL-8 production from human endothelial cells in vitro and protects mice from LPS-induced mortality and systemic inflammation & coagulopathy in vivo with good pharmacokinetics.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service