Skip to Content
Merck
All Photos(1)

Key Documents

SML1739

Sigma-Aldrich

CX08005

≥98% (HPLC)

Synonym(s):

2-(((2-(Tetradecyloxy)phenyl)amino)carbonyl)benzoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$203.00

$203.00


Estimated to ship on01 June 2025



Select a Size

Change View
5 MG
$203.00

About This Item

Empirical Formula (Hill Notation):
C28H39NO4
CAS Number:
Molecular Weight:
453.61
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$203.00


Estimated to ship on01 June 2025


Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(O)C1=C(C(NC2=C(OCCCCCCCCCCCCCC)C=CC=C2)=O)C=CC=C1

InChI key

WOEDXTWDPRNPMZ-UHFFFAOYSA-N

Biochem/physiol Actions

CX08005 is a cell-permeable benzoic acid based compound, potent and substrate-competitive PTP1B/TCPTP dual inhibitor with anti-diabetic efficacy both in vitro and in vivo. CX08005 targets protein tyrosine phosphatase 1B (PTP1B) catalytic P-loop and acts as a substrate-competitive inhibitor against PTP1B and TCPTP (IC50 = 781 and 475 nM, respectively) with much reduced potency toward LAR, SHP1, and VHR (by 52%, 89%, and 60%, respectively, at 10 μM). CX08005 enhances insulin-dependent cellular signaling (Eff. conc. 10 nM; HepG2 cells) and glucose uptake (Eff. conc. 100-500 nM; 3T3-L1 adipocityes and C2C12 myoblasts) in vitro, as well as improves glucose tolerance and insulin sensitivity in diet-induced obesity (DIO) mice and KKAy type 2 diabetes mice (50-200 mg/kg/day i.p.) in vivo with good pharmacokinetics in SD rats (Cmax = 7425.9 ng/mL, Tmax = 6.67 h,  t1/2 = 2.5 h).
CX08005 is a cell-permeable, potent and substrate-competitive PTP1B/TCPTP dual inhibitor with anti-diabetic efficacy both in vitro and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service