Skip to Content
Merck
All Photos(1)

Documents

SML1727

Sigma-Aldrich

EB1089

≥98% (HPLC)

Synonym(s):

(1R,3S,5Z)-5-[(2E)-2-[(1R,3aS,7aR)-1-[(1R,2E,4E)-6-Ethyl-6-hydroxy-1-methyl-2,4-octadien-1-yl]octahydro-7a-methyl-4H-inden-4-ylidene]et

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H46O3
CAS Number:
Molecular Weight:
454.68
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

C[C@@]12[C@](CC[C@]2([H])[C@@H](/C=C/C=C/C(CC)(O)CC)C)([H])/C(CCC1)=C/C=C3C[C@@H](O)C[C@H](O)C/3=C

InChI

1S/C30H46O3/c1-6-30(33,7-2)18-9-8-11-21(3)26-15-16-27-23(12-10-17-29(26,27)5)13-14-24-19-25(31)20-28(32)22(24)4/h8-9,11,13-14,18,21,25-28,31-33H,4,6-7,10,12,15-17,19-20H2,1-3,5H3/b11-8+,18-9+,23-13-,24-14+/t21-,25-,26-,27+,28+,29-/m1/s1

InChI key

LVLLALCJVJNGQQ-QMWUBLQMSA-N

Biochem/physiol Actions

EB1089 is a Vitamin D analog 50 to 200 times more potent than Vitamin D in inhibiting growth and inducing differentiation with half the hypercalcemic activity. Studies have found EB1089 to be more potent than vitamin D in inhibiting growth and inducing differentiation of some cancer cell lines. EB1089 enhances radio sensitivity of H460 and A549 non-small cell lung cancer (NSCLC) cells.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service