Skip to Content
Merck
All Photos(1)

Key Documents

SML1694

Sigma-Aldrich

TVB-3166

≥98% (HPLC)

Synonym(s):

4-(1-(5-(3,4-Dimethyl-1H-pyrazol-5-yl)-2,4-dimethylbenzoyl)azetidin-3-yl)benzonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$207.00
25 MG
$824.00

$207.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$207.00
25 MG
$824.00

About This Item

Empirical Formula (Hill Notation):
C24H24N4O
CAS Number:
Molecular Weight:
384.47
UNSPSC Code:
41106300
PubChem Substance ID:
NACRES:
NA.77

$207.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CC1=C(C2=C(C)C(C)=NN2)C=C(C(N3CC(C4=CC=C(C#N)C=C4)C3)=O)C(C)=C1

InChI

1S/C24H24N4O/c1-14-9-15(2)22(10-21(14)23-16(3)17(4)26-27-23)24(29)28-12-20(13-28)19-7-5-18(11-25)6-8-19/h5-10,20H,12-13H2,1-4H3,(H,26,27)

InChI key

ICDQFUFDAFKCAX-UHFFFAOYSA-N

Biochem/physiol Actions

TVB-3166 is a cell-permeable pyrrazole derivative that inhibits cellular palmitate synthesis (IC50 <100 nM) by fatty acid synthase (FASN) keto-reductase activity in a potent and reversible manner (IC50 = 42 nM), effectively inhibiting palmitate-dependent survival of various cancer cells in cultures and xenografted tumor growth in mice vivo (60-100 mg/kg/d p.o.), while exhibiting little or no anti-proliferation activity toward non-palmitate-dependent growth of cancer and non-cancer cultures. FASN inhibition by TVB-3166 treatment is also effective against the replications respiratory viruses both in cultures (Effc. conc. 200 nM against RSV and PIV3 in A549, HEp2, and HeLa cultures) and in mice (50 mg/kg/12 h, p.o.; RSV) in vivo without significant adverse effects to the host cells or mice.
TVB-3166 is a cell-permeable pyrrazole derivative that inhibits cellular palmitate synthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barbara Schroeder et al.
Cell death & disease, 12(11), 977-977 (2021-10-23)
Inhibitors of the lipogenic enzyme fatty acid synthase (FASN) have attracted much attention in the last decade as potential targeted cancer therapies. However, little is known about the molecular determinants of cancer cell sensitivity to FASN inhibitors (FASNis), which is
Jiajun Du et al.
Nature communications, 11(1), 4830-4830 (2020-09-26)
Non-invasively probing metabolites within single live cells is highly desired but challenging. Here we utilize Raman spectro-microscopy for spatial mapping of metabolites within single cells, with the specific goal of identifying druggable metabolic susceptibilities from a series of patient-derived melanoma

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service