Skip to Content
Merck
All Photos(1)

Key Documents

SML1663

Sigma-Aldrich

Butoconazole nitrate

≥90% (HPLC)

Synonym(s):

1-[4-(4-Chlorophenyl)-2-(2,6-dichlorophenyl)sulfanylbutyl]imidazole,nitric acid, Exelgyn, Femstat One, Gynazole-1, Gynomyk, (±)-1-[4-(4-Chlorophenyl)-2-[(2,6-dichlorophenyl)thio]butyl]-1H-imidazole mononitrate

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$185.00

$185.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$185.00

About This Item

Empirical Formula (Hill Notation):
C19H17Cl3N2S · HNO3
CAS Number:
Molecular Weight:
474.79
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$185.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥90% (HPLC)

form

powder

solubility

DMSO or DMF: soluble
methanol: soluble

storage temp.

2-8°C

SMILES string

ClC1=C(C(Cl)=CC=C1)SC(CCC2=CC=C(Cl)C=C2)CN3C=CN=C3.O[N+]([O-])=O

InChI

1S/C19H17Cl3N2S.HNO3/c20-15-7-4-14(5-8-15)6-9-16(12-24-11-10-23-13-24)25-19-17(21)2-1-3-18(19)22;2-1(3)4/h1-5,7-8,10-11,13,16H,6,9,12H2;(H,2,3,4)

InChI key

ZHPWRQIPPNZNML-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Butoconazole is an imidazole derivative with in vitro antifungal activity against Candida spp. In addition, Butoconazole is clinically effective against vaginal infections.[1][2][3][4] The exact mechanism for its activity is unknown. However, Butoconazole was found to interfere with the biosynthesis of lipid and ergosterol.[5]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jacobson, J.B., et al.
Acta Obstetricia et Gynecologica Scandinavica, 64, 241-244 (1985)
Odds, F.C., et al.
The Journal of Antimicrobial Chemotherapy, 14, 105-114 (1984)
Walker, K.A.M., et al.
Journal of Medicinal Chemistry, 21, 840-843 (1978)
Droegemuller, W., et al.
Obstetrics and Gynecology, 64, 530-534 (1984)
Tatum, D.M., et al.
Infection Control and Hospital Epidemiology : The Official Journal of the Society of Hospital Epidemiologists of America, 9, 122-124 (1988)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service