Skip to Content
Merck
All Photos(1)

Key Documents

SML1636

Sigma-Aldrich

Pipecuronium bromide

≥98% (HPLC)

Synonym(s):

Pipecurium bromide, RGH-1106

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$166.00
25 MG
$658.00

$166.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$166.00
25 MG
$658.00

About This Item

Empirical Formula (Hill Notation):
C35H62Br2N4O4
CAS Number:
Molecular Weight:
762.70
UNSPSC Code:
12352200
NACRES:
NA.77

$166.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

room temp

SMILES string

[Br-].[Br-].[N+]1(CCN(CC1)[C@@H]2[C@@H]([C@@]3([C@H]([C@H]4[C@@H]([C@@]5([C@H](C[C@@H]([C@H](C5)N6CC[N+](CC6)(C)C)OC(=O)C)CC4)C)CC3)C2)C)OC(=O)C)(C)C

InChI

1S/C35H62N4O4.2BrH/c1-24(40)42-32-21-26-9-10-27-28(35(26,4)23-31(32)37-15-19-39(7,8)20-16-37)11-12-34(3)29(27)22-30(33(34)43-25(2)41)36-13-17-38(5,6)18-14-36;;/h26-33H,9-23H2,1-8H3;2*1H/q+2;;/p-2/t26-,27+,28-,29-,30-,31-,32-,33-,34-,35-;;/m0../s1

InChI key

TXWBOBJCRVVBJF-YTGGZNJNSA-L

Biochem/physiol Actions

Pipecuronium is a bisquaternary aminosteroid muscle relaxant which acts at the nicotinic acetylcholine receptor of the neuromuscular junction. Pipecuronium is a non-depolarizing neuromuscular blocking agent.
Pipecuronium is a bisquaternary aminosteroid muscle relaxant.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service