Skip to Content
Merck
All Photos(1)

Key Documents

SML1618

Sigma-Aldrich

Garenoxacin mesylate monohydrate

≥98% (HPLC)

Synonym(s):

1-Cyclopropyl-8-(difluoromethoxy)-7-[(1R)-2,3-dihydro-1-methyl-1H-isoindol-5-yl]-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid methane sulfonic acid salt monohydrate, BMS 284756 mesylate, Ganefloxacin mesylate, Garenfloxacin mesylate, T-3811 mesylate

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$166.00
50 MG
$658.00

$166.00


Estimated to ship on26 April 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$166.00
50 MG
$658.00

About This Item

Empirical Formula (Hill Notation):
C23H20F2N2O4 · CH4O3S · H2O
CAS Number:
Molecular Weight:
540.53
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$166.00


Estimated to ship on26 April 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 10 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

OC(C1=CN(C2CC2)C3=C(OC(F)F)C(C4=CC(CN[C@@H]5C)=C5C=C4)=CC=C3C1=O)=O.CS(=O)(O)=O.[H]O[H]

InChI

1S/C23H20F2N2O4.CH4O3S.H2O/c1-11-15-5-2-12(8-13(15)9-26-11)16-6-7-17-19(21(16)31-23(24)25)27(14-3-4-14)10-18(20(17)28)22(29)30;1-5(2,3)4;/h2,5-8,10-11,14,23,26H,3-4,9H2,1H3,(H,29,30);1H3,(H,2,3,4);1H2/t11-;;/m1../s1

InChI key

IGTHEWGRXUAFKF-NVJADKKVSA-N

Biochem/physiol Actions

Broad spectrum fluoroquinolone antibiotic suitable for treatment of gram-positive and gram-negative bacterial infections
Garenoxacin is a broad spectrum fluoroquinolone antibiotic suitable for treatment of gram-positive and gram-negative bacterial infections. Garenoxacin has a low mutant prevention concentration and a narrow mutant selection window. Data shows that garenoxacin has anti-inflammatory properties. It significantly alleviates LPS-induced IL-8 production through the negative regulation of the ERK1/2 pathway both in the human airway epithelial cell line A549 and the monocyte cell line THP-1.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service