Skip to Content
Merck
All Photos(1)

Key Documents

SML1605

Sigma-Aldrich

OTX015

≥98% (HPLC)

Synonym(s):

6H-Thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepine-6-acetamide, 4-(4-chlorophenyl)-N-(4-hydroxyphenyl)-2,3,9-trimethyl-, (6S)-, OTX-015

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$371.00
25 MG
$1,450.00

$371.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$371.00
25 MG
$1,450.00

About This Item

Empirical Formula (Hill Notation):
C25H22ClN5O2S
CAS Number:
Molecular Weight:
491.99
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$371.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC(C=C1)=CC=C1O)C[C@H]2C3=NN=C(C)N3C(SC(C)=C4C)=C4C(C5=CC=C(Cl)C=C5)=N2

InChI

1S/C25H22ClN5O2S/c1-13-14(2)34-25-22(13)23(16-4-6-17(26)7-5-16)28-20(24-30-29-15(3)31(24)25)12-21(33)27-18-8-10-19(32)11-9-18/h4-11,20,32H,12H2,1-3H3,(H,27,33)/t20-/m0/s1

InChI key

GNMUEVRJHCWKTO-FQEVSTJZSA-N

Gene Information

Biochem/physiol Actions

It binds to bromodomain and extra-terminal domain (BET) proteins and inhibits their binding to the chromatin. This in turn prevents gene transcription. OTX015 has been shown to inhibit proliferation of cells in haematological malignancies.[1]
OTX015 is a potent inhibitor of the BET bromodomain proteins 2, 3, and 4 (BRD2/3/4).
OTX015 is a potent inhibitor of the BET bromodomain proteins 2, 3, and 4.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bromodomain inhibitor OTX015 in patients with acute leukaemia: a dose-escalation, phase 1 study.
Berthon C
The Lancet. Haematology, 3(4) (2016)
Yaping Wu et al.
Theranostics, 9(6), 1777-1793 (2019-05-01)
The bromodomain and extraterminal family members are epigenetic readers and transcriptional coactivators which are critically involved in various biological processes including tumorigenesis. BRD4 has been increasingly appreciated as a key oncogene and promising anticancer target. Here, we sought to characterize
Nana Chen et al.
Cell reports, 39(11), 110970-110970 (2022-06-16)
Analysis of The Cancer Genome Atlas and other published data of head and neck squamous cell carcinoma (HNSCC) reveals somatic alterations of the Hippo-YAP pathway in approximately 50% of HNSCC. Better strategies to target the YAP1 transcriptional complex are sought.
Leiming Wang et al.
The Journal of clinical investigation, 130(4), 1782-1792 (2019-12-25)
Recent findings have shown that inhibitors targeting bromodomain and extraterminal domain (BET) proteins, such as the small molecule JQ1, are potent growth inhibitors of many cancers and hold promise for cancer therapy. However, some reports have also revealed that JQ1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service