Skip to Content
Merck
All Photos(1)

Key Documents

SML1602

Sigma-Aldrich

RQ-00203078

≥98% (HPLC)

Synonym(s):

4-(N-(3-Chloro-5-(trifluoromethyl)pyridin-2-yl)-N-(4-(trifluoromethoxy)benzyl)sulfamoyl)benzoic acid, 4-[[[3-Chloro-5-(trifluoromethyl)-2-pyridinyl][[4-(trifluoromethoxy)phenyl]methyl]amino]sulfonyl]-benzoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$226.00
25 MG
$906.00

$226.00


Check Cart for Availability


Select a Size

Change View
5 MG
$226.00
25 MG
$906.00

About This Item

Empirical Formula (Hill Notation):
C21H13ClF6N2O5S
CAS Number:
Molecular Weight:
554.85
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$226.00


Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 25 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC1=CC(C(F)(F)F)=CN=C1N(S(C2=CC=C(C(O)=O)C=C2)(=O)=O)CC3=CC=C(OC(F)(F)F)C=C3

InChI

1S/C21H13ClF6N2O5S/c22-17-9-14(20(23,24)25)10-29-18(17)30(11-12-1-5-15(6-2-12)35-21(26,27)28)36(33,34)16-7-3-13(4-8-16)19(31)32/h1-10H,11H2,(H,31,32)

InChI key

IJGQFZYYEHCCIZ-UHFFFAOYSA-N

Biochem/physiol Actions

RQ-00203078 is an orally available potent and selective TRPM8 antagonist that potently reduces cation currents induced by menthol and other TRPM8 agonists. RQ-00203078 inhibits HSC3 and HSC4 oral squamous carcinoma cell migration and invasion in vitro.
RQ-00203078 is an orally available potent and selective TRPM8 antagonist.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alec R Nickolls et al.
Cell reports, 30(3), 932-946 (2020-01-23)
Efficient and homogeneous in vitro generation of peripheral sensory neurons may provide a framework for novel drug screening platforms and disease models of touch and pain. We discover that, by overexpressing NGN2 and BRN3A, human pluripotent stem cells can be transcriptionally

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service