Skip to Content
Merck
All Photos(1)

Key Documents

SML1601

Sigma-Aldrich

NSC59984

≥98% (HPLC)

Synonym(s):

(E)-1-(4-Methylpiperazin-1-yl)-3-(5-nitrofuran-2yl)prop-2-en-1-one, 1-(4-Methyl-1-piperazinyl)-3-(5-nitro-2-furanyl)-2-propen-1-one, 1-Methyl-4-[3-(5-nitro-2-furanyl)-1-oxo-2-propenyl]-piperazine, CID 5356520, MLS000737358

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$141.00

$141.00


Estimated to ship on26 May 2025



Select a Size

Change View
5 MG
$141.00

About This Item

Empirical Formula (Hill Notation):
C12H15N3O4
CAS Number:
Molecular Weight:
265.27
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$141.00


Estimated to ship on26 May 2025


Quality Level

Assay

≥98% (HPLC)

form

powder

color

yellow to brown

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CN1CCN(C(/C=C/C2=CC=C([N+]([O-])=O)O2)=O)CC1

InChI

1S/C12H15N3O4/c1-13-6-8-14(9-7-13)11(16)4-2-10-3-5-12(19-10)15(17)18/h2-5H,6-9H2,1H3/b4-2+

InChI key

QKTRIGNWBRHBFV-DUXPYHPUSA-N

Biochem/physiol Actions

NSC59984 targets various mutant p53 to restore wild-type p53 pathway via the activation of p73 in cancer cells.
NSC59984 targets various mutant p53 to restore wild-type p53 pathway via the activation of p73 in cancer cells. NSC59984 depletes a gain of function (GOF p53 mutants. NSC59984 is not cytotoxic against normal cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service