Skip to Content
Merck
All Photos(1)

Key Documents

SML1567

Sigma-Aldrich

MCB-613

≥98% (HPLC)

Synonym(s):

4-Ethyl-2,6-bis(pyridin-3-ylmethylene)cyclohexanone

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$141.00
25 MG
$559.00

$141.00


Check Cart for Availability


Select a Size

Change View
5 MG
$141.00
25 MG
$559.00

About This Item

Empirical Formula (Hill Notation):
C20H20N2O
CAS Number:
Molecular Weight:
304.39
UNSPSC Code:
51111800
NACRES:
NA.77

$141.00


Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

General description

MCB-613 (4-Ethyl-2,6-bis-pyridin-3-ylmethylene-cyclohexanone) belongs to the chalcone class of compounds.[1] It is an anti-neoplastic compound.[2]

Application

MCB-613 has been used to study the molecular mechanism of MCB-613-induced cell death.[2]

Biochem/physiol Actions

MCB-613 is a pan-Steroid Receptor Coactivator (SRC) stimulator. There are three steroid receptor coactivators (SRC) that interact with transcription factors and whose overexpression is assciated with a number of cancers, particularly breast cancer. MCB-613 overstimulates SRC activity in cancer cells resulting in excessive generation of reactive oxygen species (ROS), leading to cell stress and death by a process called paraptosis. MCB-613 is a cytotoxic molecule which can kill various human cancer cell lines, like HepG2 (liver), PC-3 (prostate) and MCF-7 (breast) cells.
MCB-613 is capable of inducing the transcriptional activity of steroid receptor coactivators (SRCs).[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyperactivation of nuclear receptor coactivators induces PERK-dependent cell death
Hossain MM, et al.
Testing, 9(14), 11707-11707 (2018)
Lei Wang et al.
Cancer cell, 28(2), 240-252 (2015-08-13)
By integrating growth pathways on which cancer cells rely, steroid receptor coactivators (SRC-1, SRC-2, and SRC-3) represent emerging targets in cancer therapeutics. High-throughput screening for SRC small molecule inhibitors (SMIs) uncovered MCB-613 as a potent SRC small molecule "stimulator" (SMS).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service