Skip to Content
Merck
All Photos(1)

Key Documents

SML1565

Sigma-Aldrich

A-196

≥98% (HPLC)

Synonym(s):

Cyclopentyl-(6,7-dichloro-4-pyridin-4-yl-phthalazin-1-yl)-amine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$186.00
25 MG
$741.00

$186.00


Estimated to ship on14 April 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$186.00
25 MG
$741.00

About This Item

Empirical Formula (Hill Notation):
C18H16Cl2N4
CAS Number:
Molecular Weight:
359.25
MDL number:
UNSPSC Code:
12171501
PubChem Substance ID:
NACRES:
NA.77

$186.00


Estimated to ship on14 April 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

ClC1=C(Cl)C=C(C(C2=CC=NC=C2)=NN=C3NC4CCCC4)C3=C1

InChI

1S/C18H16Cl2N4/c19-15-9-13-14(10-16(15)20)18(22-12-3-1-2-4-12)24-23-17(13)11-5-7-21-8-6-11/h5-10,12H,1-4H2,(H,22,24)

InChI key

ABGOSOMRWSYAOB-UHFFFAOYSA-N

Biochem/physiol Actions

A-196 is a potent and selective chemical inhibitor of SUV420H1 and SUV420H2 that inhibits the di- and trimethylation of H4K20me in multiple cell lines. For full characterization details, please visit the A-196 probe summary on the Structural Genomics Consortium (SGC) website.

SGC2043 is the negative control for the active probe, A-196. To request a sample of the negative control from the SGC, click here.

To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc
A-196 is a potent and selective chemical inhibitor of SUV420H1 and SUV420H2.

Features and Benefits

A-196 is an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC.
This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin Cui et al.
JCI insight, 9(11) (2024-05-07)
Activation of brown adipose tissue (BAT) thermogenesis increases energy expenditure and alleviates obesity. Here we discover that histone methyltransferase suppressor of variegation 4-20 homolog 2 (Suv420h2) expression parallels that of Ucp1 in brown and beige adipocytes and that Suv420h2 knockdown
Wendan Ren et al.
Nature communications, 12(1), 2490-2490 (2021-05-05)
DNA methylation and trimethylated histone H4 Lysine 20 (H4K20me3) constitute two important heterochromatin-enriched marks that frequently cooperate in silencing repetitive elements of the mammalian genome. However, it remains elusive how these two chromatin modifications crosstalk. Here, we report that DNA
Alena Svobodová Kovaříková et al.
Cells, 9(2) (2020-02-09)
The DNA damage response is mediated by both DNA repair proteins and epigenetic markers. Here, we observe that N6-methyladenosine (m6A), a mark of the epitranscriptome, was common in RNAs accumulated at UV-damaged chromatin; however, inhibitors of RNA polymerases I and
Dalia Rosano et al.
Cancer discovery, 14(5), 866-889 (2024-03-26)
Patients with estrogen receptor-positive breast cancer receive adjuvant endocrine therapies (ET) that delay relapse by targeting clinically undetectable micrometastatic deposits. Yet, up to 50% of patients relapse even decades after surgery through unknown mechanisms likely involving dormancy. To investigate genetic
Lenka Stixová et al.
Chromosome research : an international journal on the molecular, supramolecular and evolutionary aspects of chromosome biology, 27(1-2), 41-55 (2019-01-06)
Repair of ribosomal DNA (rDNA) is a very important nuclear process due to the most active transcription of ribosomal genes. Proper repair of rDNA is required for physiological biogenesis of ribosomes. Here, we analyzed the epigenetics of the DNA damage

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service