Skip to Content
Merck
All Photos(1)

Key Documents

SML1560

Sigma-Aldrich

Azoramide

≥98% (HPLC)

Synonym(s):

N-[2-[2-(4-Chlorophenyl)-4-thiazolyl]ethyl]-butanamide, N-{2-[2-(4-Chlorophenyl)-1,3-thiazol-4-yl]ethyl} butanamide

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$167.00
50 MG
$577.00

$167.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$167.00
50 MG
$577.00

About This Item

Empirical Formula (Hill Notation):
C15H17ClN2OS
CAS Number:
Molecular Weight:
308.83
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$167.00


Estimated to ship on18 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 30 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC1=CC=C(C2=NC(CCNC(CCC)=O)=CS2)C=C1

InChI

1S/C15H17ClN2OS/c1-2-3-14(19)17-9-8-13-10-20-15(18-13)11-4-6-12(16)7-5-11/h4-7,10H,2-3,8-9H2,1H3,(H,17,19)

InChI key

VYBFWKKCWTXCQX-UHFFFAOYSA-N

Biochem/physiol Actions

Azoramide is a modulator of the unfolded protein response with antidiabetic activity. It has dual activity: Azoramide improves endoplasmic reticulum (ER) protein-folding ability and stimulates the expression of ER chaperones that protect cells against ER stress. Azoramide was shown to protect cells from death and dysfunction in multiple models of ER stress. It showed antidiabetic activity in two independent mouse models of obesity by improving insulin sensitivity and protecting pancreatic β cell function against ER stress.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service