Skip to Content
Merck
All Photos(1)

Key Documents

SML1551

Sigma-Aldrich

GV-58

≥97% (HPLC)

Synonym(s):

(2R)-2-[[6-[[(5-Methyl-2-thienyl)methyl]amino]-9-propyl-9H-purin-2-yl]amino]-1-butanol

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$109.00
5 MG
$350.00

$109.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$109.00
5 MG
$350.00

About This Item

Empirical Formula (Hill Notation):
C18H26N6OS
CAS Number:
Molecular Weight:
374.50
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$109.00


Estimated to ship on20 May 2025


Request a Bulk Order

Quality Level

Assay

≥97% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

CCCN1C2=NC(N[C@H](CC)CO)=NC(NCC3=CC=C(C)S3)=C2N=C1

InChI

1S/C18H26N6OS/c1-4-8-24-11-20-15-16(19-9-14-7-6-12(3)26-14)22-18(23-17(15)24)21-13(5-2)10-25/h6-7,11,13,25H,4-5,8-10H2,1-3H3,(H2,19,21,22,23)/t13-/m1/s1

InChI key

DPTXJOUVBMUSGY-CYBMUJFWSA-N

Biochem/physiol Actions

GV-58 is a roscovitine analog that retains the ability to prolong the open state of calcium channels, but unlike roscovitine, is inactive against Cdk activity. GV-58 is a selective agonist of N-type and P/Q type calcium channels, which are critical to the triggering of neurotransmitter release at the neuromuscular junction. GV-58 has been studied as a possible therapeutic agent in a mouse model of Lambert-Eaton myasthenic syndrome (LEMS). The EC50 values for activation of N-, P/Q- and L- type calcium channels are 6.8, 9.9 and >100 μM, respectively.
GV-58 is a selective agonist of N-type and P/Q type calcium channels.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brett J Hilton et al.
Neuron, 110(1), 51-69 (2021-10-28)
Axons in the adult mammalian central nervous system fail to regenerate after spinal cord injury. Neurons lose their capacity to regenerate during development, but the intracellular processes underlying this loss are unclear. We found that critical components of the presynaptic
Bryan J Black et al.
Frontiers in cellular neuroscience, 11, 304-304 (2017-10-17)
Motor neurons are the site of action for several neurological disorders and paralytic toxins, with cell bodies located in the ventral horn (VH) of the spinal cord along with interneurons and support cells. Microelectrode arrays (MEAs) have emerged as a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service