Skip to Content
Merck
All Photos(1)

Key Documents

SML1544

Sigma-Aldrich

VPC-14228

≥98% (HPLC)

Synonym(s):

2-Morpholino-4-phenylthiazole, 4-(4-Phenyl-2-thiazolyl)-morpholine, 4-(4-Phenylthiazol-2-yl)morpholine

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$559.00

$559.00


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$559.00

About This Item

Empirical Formula (Hill Notation):
C13H14N2OS
CAS Number:
Molecular Weight:
246.33
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

$559.00


Estimated to ship on16 April 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

C1(N2CCOCC2)=NC(C3=CC=CC=C3)=CS1

InChI

1S/C13H14N2OS/c1-2-4-11(5-3-1)12-10-17-13(14-12)15-6-8-16-9-7-15/h1-5,10H,6-9H2

InChI key

MRNNJVNMXOZEQC-UHFFFAOYSA-N

Biochem/physiol Actions

VPC-14228 is a potent and specific inhibitor of human androgen receptor (AR) that binds to AR DNA-binding domain (AR DBD). Apparently VPC-14228 blocks the transcriptional activity of constitutively active AR splice variants.
VPC-14228 is a potent and specific inhibitor of human androgen receptor (AR).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service