Skip to Content
Merck
All Photos(1)

Key Documents

SML1449

Sigma-Aldrich

G3335

≥98% (HPLC)

Synonym(s):

H-Trp-Glu-OH, L-Tryptophyl-L-glutamic acid, WE

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$141.00
250 MG
$559.00

$141.00


Check Cart for Availability


Select a Size

Change View
50 MG
$141.00
250 MG
$559.00

About This Item

Empirical Formula (Hill Notation):
C16H19N3O5
CAS Number:
Molecular Weight:
333.34
UNSPSC Code:
12352200
NACRES:
NA.77

$141.00


Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

storage temp.

−20°C

SMILES string

[nH]1c2c(c(c1)C[C@H](N)C(=O)N[C@@H](CCC(=O)O)C(=O)O)cccc2

InChI

1S/C16H19N3O5/c17-11(7-9-8-18-12-4-2-1-3-10(9)12)15(22)19-13(16(23)24)5-6-14(20)21/h1-4,8,11,13,18H,5-7,17H2,(H,19,22)(H,20,21)(H,23,24)/t11-,13-/m0/s1

InChI key

PWIQCLSQVQBOQV-AAEUAGOBSA-N

Biochem/physiol Actions

G3335 ( L-tryptophyl-L-glutamic acid) is a cell-permeable dipeptide that acts as a selective and reversible PPARγ antagonist with a Kd value of 8.34 μM for PPARγ and much lower activity at other PPAR subtypes. G3335 reversibly and competitively blocks activation of PPARγ by the PPARγ agonist rosiglitazone, and conversely, in rat astrocyte culture higher levels of rosiglitazone reduced harmful effects induced by G3335. At higher concentration G3335 may act as a PPARα agonist, interacting with the PPARα ligand binding domain with a KD of 120 μM. G3335 is reported to reduce hepatic lipid accumulation in lipid-loaded hepatocytes by activation of PPARα.
G3335 ( L-tryptophyl-L-glutamic acid) is a cell-permeable dipeptide that acts as a selective and reversible PPARγ antagonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service