Skip to Content
Merck
All Photos(1)

Key Documents

SML1443

Sigma-Aldrich

L48H37

≥98% (HPLC)

Synonym(s):

(3E,5E)-1-ethyl-3,5-bis[(2,3,4-trimethoxyphenyl)methylidene]piperidin-4-one, 1-Ethyl-3,5-bis(3,4,5-trimethoxybenzylidene)piperidin-4-one, 1-Ethyl-3,5-bis[(2,3,4-trimethoxyphenyl)methylene]-4-piperidinone

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$141.00
25 MG
$559.00

$141.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$141.00
25 MG
$559.00

About This Item

Empirical Formula (Hill Notation):
C27H33NO7
CAS Number:
Molecular Weight:
483.55
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$141.00


Estimated to ship on25 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O=C(/C(CN(CC)C/1)=C/C2=C(OC)C(OC)=C(OC)C=C2)C1=C\C3=CC=C(OC)C(OC)=C3OC

InChI

1S/C27H33NO7/c1-8-28-15-19(13-17-9-11-21(30-2)26(34-6)24(17)32-4)23(29)20(16-28)14-18-10-12-22(31-3)27(35-7)25(18)33-5/h9-14H,8,15-16H2,1-7H3/b19-13+,20-14+

InChI key

XLPNFINERWLGNC-IWGRKNQJSA-N

Application

L48H37 has been used as a myeloid differentiator protein 2 (MD2) antagonist/inhibitor:
  • to investigate intracellular signaling of LPS in signal transduction pathways via endosomic toll-like receptor 4 (TLR4) in colonic epithelium[1]
  • to determine its anti-cancer effects in pancreatic ductal adenocarcinoma (PDAC)[2]
  • to study its effects on lipopolysaccharide (LPS) and palmitate CC chemokine ligand 2 (CCL2) responses in normal kidney proximal tubule cells[3]

Biochem/physiol Actions

L48H37 can stimulate apoptosis and prevent proliferation in pancreatic cancer cells, especially in the absence of histone-lysine N-methyltransferase 2D (KMT2D). It has an unsaturated monoketone structure, which helps to enhance its stability and anti-cancer effects. L48H37 may possess an anti-neoplastic effect on human pancreatic ductal adenocarcinoma (PDAC) cells.[2]
L48H37 is a potent and chemically stable anti-inflammatory curcumin analog that inhibits LPS-induced inflammation through the myeloid differentiation 2 (MD-2) and toll-like receptor 4 (TLR4) complex. L48H37 is a potent MD2 inhibitor that binds to the hydrophobic region of MD-2 and blocks the interaction between MD2 and LPS. L48H37 significantly improves survival and protected lung injury in LPS-induced septic mice.
L48H37 is a potent and chemically stable anti-inflammatory curcumin analog.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Si-Si Li et al.
World journal of gastrointestinal oncology, 11(8), 599-621 (2019-08-23)
Novel therapeutic strategies are urgently needed for patients with a delayed diagnosis of pancreatic ductal adenocarcinoma (PDAC) in order to improve their chances of survival. Recent studies have shown potent anti-neoplastic effects of curcumin and its analogues. In addition, the
Charles C Okechukwu et al.
Peptides, 136, 170436-170436 (2020-11-13)
The renin-angiotensin system (RAS) plays a critical role in the regulation of blood pressure. Inappropriate activation of the RAS, particularly stimulation of the ACE-Ang II-AT1 receptor axis is a key factor in hypertension and AT1R antagonists (ARBs) are first line
Ravi Holani et al.
Gut microbes, 12(1), 1785802-1785802 (2020-07-14)
We hypothesized that the antimicrobial peptide cathelicidin has a physiological role in regulating gut inflammatory homeostasis. We determined that cathelicidin synergizes with LPS to facilitate its internalization and signaling via endosomic TLR4 in colonic epithelium, evoking synthesis of the human

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service