Skip to Content
Merck
All Photos(1)

Key Documents

SML1432

Sigma-Aldrich

PHCCC

≥97% (HPLC)

Synonym(s):

7,7a-Dihydro-7-(hydroxyimino)-N-phenyl-benzo[b]cyclopropa[e]pyran-1a(1H)-carboxamide, N-Phenyl-7-(hydroxyimino)cyclopropa[b]chromen-1a-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$494.00

$494.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$494.00

About This Item

Empirical Formula (Hill Notation):
C17H14N2O3
CAS Number:
Molecular Weight:
294.30
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

$494.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC1=CC=CC=C1)C2(C/3C2)OC4=C(C=CC=C4)C3=N/O

InChI

1S/C17H14N2O3/c20-16(18-11-6-2-1-3-7-11)17-10-13(17)15(19-21)12-8-4-5-9-14(12)22-17/h1-9,13,21H,10H2,(H,18,20)/b19-15+

InChI key

FPXPIEZPAXSELW-XDJHFCHBSA-N

Biochem/physiol Actions

PHCCC is a positive allosteric modulator (PAM) at the mGluR4 subtype that exhibits anxiolytic and anti-Parkinsonian effects in animal models. PHCCC is a potent group I metabotropic glutamate receptor antagonist.
PHCCC is a positive allosteric modulator (PAM) at the mGluR4 subtype. PHCCC is a potent group I metabotropic glutamate receptor antagonist.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service