Skip to Content
Merck
All Photos(1)

Key Documents

SML1422

Sigma-Aldrich

CCG-203971

≥98% (HPLC)

Synonym(s):

N-(4-Chlorophenyl)-1-[3-(2-furanyl)benzoyl]-3-piperidinecarboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$128.00
25 MG
$412.00

$128.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$128.00
25 MG
$412.00

About This Item

Empirical Formula (Hill Notation):
C23H21ClN2O3
CAS Number:
Molecular Weight:
408.88
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$128.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

storage temp.

2-8°C

SMILES string

ClC(C=C1)=CC=C1NC(C(C2)CCCN2C(C3=CC(C4=CC=CO4)=CC=C3)=O)=O

InChI

1S/C23H21ClN2O3/c24-19-8-10-20(11-9-19)25-22(27)18-6-2-12-26(15-18)23(28)17-5-1-4-16(14-17)21-7-3-13-29-21/h1,3-5,7-11,13-14,18H,2,6,12,15H2,(H,25,27)

InChI key

HERLZBNILRVHQN-UHFFFAOYSA-N

Biochem/physiol Actions

CCG-203971 is an inhibitor of the Rho/MKL1/SRF transcriptional pathway, which has been shown to play a role in metastasis of melanoma and breast cancer and clinically associated with castration-resistant prostate cancer. CCG-203971 is a second-generation analog of CCG-1423 (SML0987) with an IC50 of 4.2 μM vs 1 μM for CCG-1423, but less cytotoxicity. In mouse studies, CCG-203971 inhibited invasion of PC-3 prostate cancer cells and was well tolerated up to doses of 100 mg/kg IP over 5 days.

The Rho/MRTF/SRF pathway has also been shown to be involved in multiple types of solid organ fibrosis. CCG-203971 repressed both matrix-stiffness and TGF-β-mediated fibrogenesis in human colonic myofibroblasts and showed antifibrotic activity in a murine model of skin injury and in pulmonary fibrosis lung fibroblasts.
CCG-203971 is an inhibitor of the Rho/MKL1/SRF transcriptional pathway.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lídia Faria et al.
Disease models & mechanisms, 16(2) (2023-02-23)
Alterations in the expression or function of cell adhesion molecules have been implicated in all steps of tumor progression. Among those, P-cadherin is highly enriched in basal-like breast carcinomas, playing a central role in cancer cell self-renewal, collective cell migration
Allison P Kann et al.
Cell stem cell, 29(6), 933-947 (2022-05-22)
Many tissues harbor quiescent stem cells that are activated upon injury, subsequently proliferating and differentiating to repair tissue damage. Mechanisms by which stem cells sense injury and transition from quiescence to activation, however, remain largely unknown. Resident skeletal muscle stem

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service