Skip to Content
Merck
All Photos(1)

Key Documents

SML1420

Sigma-Aldrich

FSL-1

≥80% (HPLC)

Synonym(s):

(R,S)-Pam2Cys-Gly-Asp-Pro-Lys-His-Pro-Lys-Ser-Phe-OH, Fibroblast-stimulating Lipopeptide, Pam2CGDPKHPKSF, S-[2,3-Bis[(1-Oxohexadecyl)oxy]propyl]-L-cysteinylglycyl-L-a-aspartyl-L-prolyl-L-lysyl-L-histidyl-L-prolyl-L-lysyl-L-seryl-L-phenylalanine

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$413.00

$413.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$413.00

About This Item

Empirical Formula (Hill Notation):
C84H140N14O18S
CAS Number:
Molecular Weight:
1666.16
UNSPSC Code:
12352200
NACRES:
NA.77

$413.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥80% (HPLC)

form

powder

color

white to off-white

solubility

H2O: soluble

storage temp.

−20°C

Biochem/physiol Actions

FSL-1 (Pam2CGDPKHPKSF) is a synthetic lipoprotein that acts as a Toll-like Receptor 2/6 (TLR2/6) agonist and activator of nuclear transcription factor NF-KB.
FSL-1 (Pam2CGDPKHPKSF) is a synthetic lipoprotein that acts as a Toll-like Receptor 2/6 (TLR2/6) agonist and activator of nuclear transcription factor NF-KB. FSL-1 is based on the N-terminal part of the 44-kDa lipoprotein LP44 of Mycoplasma salivarium. FSL-1 showed higher stimulatory activity than MALP-2 in its ability to induce TNF-α production in macrophages and NF-κB in TLR2/TLR6 transfected HEK293 cells.
FSL-1 is a bacterial-derived toll-like receptor 2/6 agonist. It is capable of increasing resistance to herpes simplex virus type 2 (HSV-2) infection.[1]

Reconstitution

Reconstitute in endotoxin-free water.

Other Notes

Air sensitive

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

FSL-1, a bacterial-derived toll-like receptor 2/6 agonist, enhances resistance to experimental HSV-2 infection
Rose WA, et al.
Virology Journal, 6, 195-195 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service